Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Recommended positive controls: HeLa, COL3A1 shRNA-transfected HeLa, SK-N-SH, SK-N-AS, Neuro2A, NIH-3T3, BCL-1, Raw264.7, C2C12, Mouse ESC, PC-12, Rat2, rat liver.
Predicted reactivity: Mouse (92%), Rat (92%), Xenopus laevis (80%), Pig (94%), Chicken (83%), Rhesus Monkey (98%), Bovine (94%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Collagen III is a fibrillar collagen that is found in extensible connective tissues such as skin, lung, and the vascular system, frequently in association with type I Collagen. Mutations in th gene are associated with Ehlers Danlos syndrome type IV, and with aortic and arterial aneurysms. Although alternate transcripts have been detected for this gene, they are the result of mutations; these mutations alter splicing, often leading to the exclusion of multiple exons.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-1 type III collagen; alpha-1(III) procollagen; alpha1 (III) collagen; COL 3; COL3; COL3 alpha 1; Collagen 3; Collagen alpha-1(III) chain; collagen, fetal; collagen, type III, alpha 1; Collagen3; Ehlers-Danlos syndrome type IV, autosomal dominant; FLJ34534; MGC93704; procollagen, type III, alpha 1; unnamed protein product
Gene Aliases: COL3A1; EDS4A; EDSVASC; PMGEDSV
UniProt ID: (Human) P02461, (Rat) P13941
Entrez Gene ID: (Human) 1281, (Rat) 84032
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support