Positive control: skin, liver
Collagen IV is a major constituent of the basement membranes along with laminins and enactins. Collagen IV is composed of alpha 1 IV chain and alpha 2 IV chain in 2:1 ratio. Collagen IV can form insoluble fibers with high tensile strength. Antibody to collagen IV is useful in detecting the loss of parts of basement membrane in carcinomas.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha1(IV) collagen; Arresten; Canstatin; col4a3; col4a4; Collagen 4; Collagen alpha-1(IV) chain; Collagen alpha-2(IV) chain; Collagen alpha-3(IV) chain; Collagen alpha-4(IV) chain; collagen type IV alpha3 chain; Del(8)Bru44H; procollagen, type IV, alpha 1; procollagen, type IV, alpha 2; procollagen, type IV, alpha 3; procollagen, type IV, alpha 4; retinal anterior wiring; Tumstatin; Type IV Collagen
Gene Aliases: [a]3(IV); [a]4(IV); alpha3(IV); Bru; Col4a-1; Col4a-2; Col4a1; Col4a2; Col4a3; Col4a4; Del(8)44H; E130010M05Rik; Raw; Svc
UniProt ID: (Mouse) P02463, (Mouse) P08122, (Mouse) Q9QZS0, (Mouse) Q9QZR9
Entrez Gene ID: (Mouse) 12826, (Mouse) 12827, (Mouse) 12828, (Mouse) 12829
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support