Sequence of this protein is as follows: EYSKSLPGLS TSYAALLRIK KSSSSSLFGS KTRPRYSSPS LGTLSASSPS WLGAAQNYYS PINLYHSSDA FKQDESVDYG PVFVQEPDDI IFPTDSDEKK VALNCEVRGN PVPSYRWLRN GTEIDLESDY RYSLIDGTFI ISNPSEAKDS GHYQCLATNT VGSILSREAT LQFAYLGNFS GRTRSAVSVR EGQGVVLMCS PP
The protein encoded by this gene is a member of the immunoglobulin superfamily, and contactin family, which mediate cell surface interactions during nervous system development. This protein is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Contactin-5; hNB-2; MGC163491; neural adhesion molecule; Neural recognition molecule NB-2
Gene Aliases: 6720426O10Rik; A830025P08Rik; CNTN5; Gm507; HNB-2s; NB-2
UniProt ID: (Human) O94779, (Mouse) P68500, (Rat) P97527
Entrez Gene ID: (Human) 53942, (Mouse) 244682, (Rat) 114589
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support