The neural adhesion molecule Contactin-6, also known as NB-3, is a contactin/F3 subgroup member of immunoglobulin superfamily. It is expressed exclusively in the nervous system and mainly upregulated at the early postnatal stage during mouse brain development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Contactin-6; hNB-3; mNB-3; neural adhesion molecule; Neural recognition molecule NB-3
Gene Aliases: CNTN6; NB-3; NB3
UniProt ID: (Human) Q9UQ52, (Mouse) Q9JMB8
Entrez Gene ID: (Human) 27255, (Mouse) 53870
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support