Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CRY1 Antibodies

          Proteintech

          Cryptochrome 1 Recombinant Rabbit Monoclonal Antibody (240520B12)

          View all (19) CRY1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Cryptochrome 1 Recombinant Rabbit Monoclonal Antibody (240520B12)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          Cryptochrome 1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Cryptochrome 1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Cryptochrome 1 Antibody (83602-5-RR) in WB

          Various lysates were subjected to SDS PAGE followed by western blot with 83602-5-RR (CRY1 antibody) at dilution of 1:4,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Cryptochrome 1 Antibody in Western Blot (WB)
          Cryptochrome 1 Antibody in Surface plasmon resonance (SPR)

          Product Details

          83602-5-RR

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:8,000
          -

          ELISA (ELISA)

          Assay-dependent
          -

          Surface plasmon resonance (SPR)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293F

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          240520B12

          Immunogen

          Cryptochrome 1 fusion protein Ag4270

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence:
          RPRMNANSL LASPTGLSPY LRFGCLSCRL FYFKLTDLYK KVKKNSSPPL SLYGQLLWRE FFYTAATNNP RFDKMEGNPI CVQIPWDKNP EALAKWAEGR TGFPWIDAIM TQLRQEGWIH HLARHAVACF LTRGDLWISW EEGMKVFEEL LLDADWSINA GSWMWLSCSS FFQQFFHCYC PVGFGRRTDP NGDYIRRYLP VLRGFPAKYI YDPWNAPEGI QKVAKCLIGV NYPKPMVNHA EASRLNIERM KQIYQQLSRY RGLGLLASVP SNPNGNGGFM GYSAENIPGC SSSGSCSQGS GILHYAHGDS QQTHLLKQGR SSMGTGLSGG KRPSQEEDTQ SIGPKVQRQS TN

          Target Information

          Various biochemical, physiological and behavioral processes display circadian rhythms controlled by an internal biological clock. The central "gears" driving this clock appear to be composed of an autoregulatory transcription/post translation-based feedback loop. Cryptochrome 1 (CRY1) and 2 (CRY2) are DNA-binding flavoproteins that bear some homology to blue-light receptors and photolyases. In Drosophila, CRY is a photoreceptor for the circadian clock where it binds to the clock component TIM in a light-dependent fashion and blocks its function. Mammalian CRY1 and CRY2 function via light-independent interactions with circadian genes CLOCK and BMAL1, as well as with PER1, PER2, and TIM. They seem to act as light-independent components of the circadian clock and likely regulate Per1 transcriptional cycling via interactions with both the activator and its feedback inhibitors. Mutant mice not expressing the Cry1 or Cry2 protein display accelerated and delayed periodicity of locomotor activity, respectively. It appears that the combination of both proteins working together is essential to synchronize the organism to circadian phases. A critical balance between Cry1 and Cry2 is required for proper clock function; in complete darkness, double-mutant mice present with instantaneous arrhythmicity, indicating the absence of an internal circadian clock.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ATCRY1; BLU1; BLUE LIGHT PHOTORECEPTOR; BLUE LIGHT UNINHIBITED 1; cryptochrome 1 (photolyase-like); CRYPTOCHROME 1 APOPROTEIN; cryptochrome circadian clock 1; Cryptochrome-1; ELONGATED HYPOCOTYL 4; HY4; OOP2; OUT OF PHASE 2; T3H13.14; T3H13_14; unnamed protein product

          View more View less

          Gene Aliases: CRY1; DSPD; PHLL1

          View more View less

          UniProt ID: (Human) Q16526

          View more View less

          Entrez Gene ID: (Human) 1407

          View more View less

          Function(s)
          nucleotide binding DNA binding double-stranded DNA binding deoxyribodipyrimidine photo-lyase activity DNA (6-4) photolyase activity protein binding photoreceptor activity blue light photoreceptor activity ligand-dependent nuclear receptor binding kinase binding protein kinase binding phosphatase binding histone deacetylase binding E-box binding FAD binding DNA-binding transcription factor binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter gluconeogenesis regulation of gluconeogenesis circadian rhythm response to light stimulus blue light signaling pathway response to activity lipid storage negative regulation of protein ubiquitination positive regulation of protein ubiquitination response to insulin circadian regulation of gene expression response to glucagon glucose homeostasis regulation of circadian rhythm negative regulation of circadian rhythm signal transduction in response to DNA damage entrainment of circadian clock by photoperiod negative regulation of gluconeogenesis positive regulation of gluconeogenesis negative regulation of G-protein coupled receptor protein signaling pathway negative regulation of transcription, DNA-templated rhythmic process regulation of DNA damage checkpoint negative regulation of glucocorticoid receptor signaling pathway negative regulation of glucocorticoid secretion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline