Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PA1-21774 detects Cyclin A in Human, Rat and Mouse samples.
PA1-21774 has been successfully used in Immunoprecipitation and Western Blot procedures.
PA1-21774 immunogen corresponds to Synthetic peptide: C-KYHGVSLLNPPETLNL, corresponding to amino acids 417-432 of Human Cyclin A..
Cyclins are regulatory subunits of the cyclin-dependent kinases (cdks) and they control transition at different specific phases of the cell cycle. The temporal expression of cyclins is tightly regulated and subsequently plays a critical role in controlling the enzymatic activity of cdks. These cyclin / cdk complexes are essential for passage through specific stages in the cell cycle. In mammalian somatic cells, cyclin A is required for S phase and passage through G2-phase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cyclin A; Cyclin A2 (Cyclin A); Cyclin-A; Cyclin-A2
Gene Aliases: CCN1; CCNA; CCNA2; Cyca; Cyca2
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support