Immunogen sequence: PSVLALCLLN LEVETLKSVE LLEILLLVKK HSKINDTEFF YWRELVSKCL AEYSSPECCK PDLKKLVWIV SRRTAQNLHN SYYSVPELPT IPEGGCFDES ES
Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.
Cyclin G2 may play a role in growth regulation and in negative regulation of cell cycle progression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cyclin-G2
Gene Aliases: CCNG2
UniProt ID: (Human) Q16589
Entrez Gene ID: (Human) 901
Molecular Function:
enzyme modulator
kinase activator
kinase modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support