Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Recommended positive controls: A549, HeLa, HepG2, HCT116.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between 40 kDa and 68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CK-2e; Cytokeratin-2e; cytoskeletal protein; epithelial keratin 2e; Epithelial keratin-2e; K2e; keratin 2, type II; keratin 2A (epidermal ichthyosis bullosa of Siemens); keratin 2e; Keratin, type II cytoskeletal 2 epidermal; Keratin-2 epidermis; Keratin-2e; MGC116967; MGC116968; Type-II keratin Kb2
Gene Aliases: CK-2e; K2E; KRT2; KRT2A; KRT2E; KRTE
UniProt ID: (Human) P35908
Entrez Gene ID: (Human) 3849, (Dog) 444850
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support