Product References
Xp11.2 translocation renal cell carcinoma with NONO-TFE3 gene fusion: morphology, prognosis, and potential pitfall in detecting TFE3 gene rearrangement.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc
Xia QY,Wang Z,Chen N,Gan HL,Teng XD,Shi SS,Wang X,Wei X,Ye SB,Li R,Ma HH,Lu ZF,Zhou XJ,Rao Q
MA112781 was used in immunohistochemistry - paraffin section to characterize eight cases of renal cell carcinoma cases with NONO-TFE3
Wed Mar 01 00:00:00 EST 2017
Reevaluation of MAML2 fusion-negative mucoepidermoid carcinoma: a subgroup being actually hyalinizing clear cell carcinoma of the salivary gland with EWSR1 translocation.
Human pathology
Hsieh MS,Wang H,Lee YH,Ko JY,Chang YL
MA112781 was used in immunohistochemistry - paraffin section to use MAML2 staining to distinguish cases of hyalinizing clear cell carcinoma from mucoepidermoid carcinoma
Wed Mar 01 00:00:00 EST 2017
Expression of CD24 and Siglec-10 in first trimester placenta: implications for immune tolerance at the fetal-maternal interface.
Histochemistry and cell biology
Sammar M,Siwetz M,Meiri H,Fleming V,Altevogt P,Huppertz B
MA112781 was used in immunohistochemistry - paraffin section to determine if the presence of the CD24-Siglec-10 in the placental bed and maternal uterine glands suggests a possible role in mediating immune tolerance at the fetal-maternal interface
Mon May 01 00:00:00 EDT 2017
Increased utilization, verification, and clinical implications of immunocytochemistry: Experience in a northern New England hospital.
Diagnostic cytopathology
Sauter JL,Ambaye AB,Mount SL
MA1-12781 was used in immunohistochemistry to study a northern New England hospital and increased utilization, verification, and clinical implications of immunocytochemistry
Tue Sep 01 00:00:00 EDT 2015
Tumor-induced STAT3 activation in monocytic myeloid-derived suppressor cells enhances stemness and mesenchymal properties in human pancreatic cancer.
Cancer immunology, immunotherapy : CII
Panni RZ,Sanford DE,Belt BA,Mitchem JB,Worley LA,Goetz BD,Mukherjee P,Wang-Gillam A,Link DC,Denardo DG,Goedegebuure SP,Linehan DC
MA1-12781 was used in immunohistochemistry to study the mechanism by which STAT3 activation promotes stemness in pancreatic cancer cancer stem cells
Thu May 01 00:00:00 EDT 2014
New diagnostic markers in salivary gland tumors.
European archives of oto-rhino-laryngology : official journal of the European Federation of Oto-Rhino-Laryngological Societies (EUFOS) : affiliated with the German Society for Oto-Rhino-Laryngology - Head and Neck Surgery
Schneider S,Kloimstein P,Pammer J,Brannath W,Grasl MCh,Erovic BM
MA1-12781 was used in immunohistochemistry to study the expresion of tumor antigens in order to identify novel diagnostic markers of parotid gland tumors
Tue Jul 01 00:00:00 EDT 2014
Amnion-derived mesenchymal stromal cells show a mesenchymal-epithelial phenotype in culture.
Cell and tissue banking
König J,Lang I,Siwetz M,Fröhlich J,Huppertz B
MA1-12781 was used in immunohistochemistry to study the mesenchymal-epithelial phenotype displayed by cultured mesenchymal stromal cells derived from the amniotic membrane
Sun Jun 01 00:00:00 EDT 2014
Three-dimensional culture and identification of human eccrine sweat glands in matrigel basement membrane matrix.
Cell and tissue research
Li H,Chen L,Zhang M,Tang S,Fu X
MA1-12781 was used in immunohistochemistry to develop a matrigel-based system for the 3-dimensional culture of human eccrine sweat glands
Sun Dec 01 00:00:00 EST 2013
Identification of novel target proteins in sebaceous gland carcinoma.
Head & neck
Erovic BM,Al Habeeb A,Harris L,Goldstein DP,Kim D,Ghazarian D,Irish JC
MA1-12781 was used in immunohistochemistry to identify novel therapeutic target proteins in sebaceous gland carcinoma
Wed May 01 00:00:00 EDT 2013
Stromal-epithelial crosstalk provides a suitable microenvironment for the progression of ovarian cancer cells in vitro.
Cancer investigation
Fu S,Dong L,Sun W,Xu Y,Gao L,Miao Y
MA1-12781 was used in immunocytochemistry to study the in vivo progression of ovarian cancer cells and the role of stromal-epithelial interactions in creating an appropriate microenvironment
Fri Nov 01 00:00:00 EDT 2013
Specimen quality evaluation in Canadian biobanks participating in the COEUR repository.
Biopreservation and biobanking
Le Page C,Köbel M,de Ladurantaye M,Rahimi K,Madore J,Babinszky S,Bachvarov DR,Bachvarova M,Beauchamp MC,Cass CE,Chadwick D,Colleen C,Damaraju S,Dufour J,Gotlieb WH,Kalloger SE,Portelance L,McAlpine JN,Matte I,Piché A,Shaw P,Roehrl MH,Vanderhyden BC,Watson PH,Huntsman DG,Provencher DM,Mes-Masson AM
MA1-12781 was used in immunohistochemistry to study the quality of biological samples held in Canadian biobanks forming part of the COEUR repository
Mon Apr 01 00:00:00 EDT 2013
Signet-ring cell carcinoma of the breast: a case report.
World journal of surgical oncology
Li X,Feng YF,Wei WD,Liu P,Xie ZM,Wang J,Xie XM
MA1-12781 was used in immunohistochemistry - paraffin section to present the case of a 46-year-old woman with primary signet-ring cell carcinoma of the breast
Fri Aug 09 00:00:00 EDT 2013
Serrated pathway adenocarcinomas: molecular and immunohistochemical insights into their recognition.
PloS one
Gurzu S,Szentirmay Z,Toth E,Bara T,Bara T,Jung I
MA1-12781 was used in immunohistochemistry to establish criteria for the recognition of serrated pathway adenocarcinomas
Tue Aug 27 00:00:00 EDT 2013
Prognostic and predictive markers in medullary thyroid carcinoma.
Endocrine pathology
Erovic BM,Kim D,Cassol C,Goldstein DP,Irish JC,Asa SL,Mete O
MA1-12781 was used in immunohistochemistry to identify markers with progniostic and predictive value in patients with medullary thyroid cancer
Sat Dec 01 00:00:00 EST 2012
Biomarkers of parathyroid carcinoma.
Endocrine pathology
Erovic BM,Harris L,Jamali M,Goldstein DP,Irish JC,Asa SL,Mete O
MA1-12781 was used in immunohistochemistry to establish a biomarker panel suitable for the identification of parathyroid carcinomas
Sat Dec 01 00:00:00 EST 2012
Significance of TRIM29 and β-catenin expression in non-small-cell lung cancer.
Journal of the Chinese Medical Association : JCMA
Zhou ZY,Yang GY,Zhou J,Yu MH
MA1-12781 was used in immunohistochemistry to study non-small cell lung cancer and the potential clinicopathological significance of TRIM29 and beta-catenin expression
Fri Jun 01 00:00:00 EDT 2012
Adenocarcinoma with sarcomatous dedifferentiation arising from mature cystic teratoma of the anterior mediastinum.
Pathology, research and practice
Kim JY,Lee CH,Park WY,Kim JY,Kim AR,Shin N,Park DY,Huh GY
MA1-12781 was used in immunohistochemistry to report on a case of adenocarcinoma with sarcomatous dedifferentiation arising from mature cystic teratoma of the anterior mediastinum
Sat Dec 15 00:00:00 EST 2012
Aberrant pattern of the cytokeratin 7/cytokeratin 20 immunophenotype in colorectal adenocarcinomas with BRAF mutations.
Pathology, research and practice
Gurzu S,Jung I
MA1-12781 was used in immunohistochemistry to study the aberrant cytokeratin 7/cytokeratin 20 immunophenotype in colorectal adenocarcinomas with BRAF mutations
Thu Mar 15 00:00:00 EDT 2012
Miliary brain metastases from occult lung adenocarcinoma: Radiologic and histopathologic confirmation.
Journal of neurosciences in rural practice
Kahveci R,Gürer B,Kaygusuz G,Sekerci Z
MA1-12781 was used in immunohistochemistry to report on a case of miliary brain metastases from occult lung adenocarcinoma
Sat Sep 01 00:00:00 EDT 2012
A new case of primary signet-ring cell carcinoma of the cervix with prominent endometrial and myometrial involvement: Immunohistochemical and molecular studies and review of the literature.
World journal of surgical oncology
Giordano G,Pizzi S,Berretta R,D'Adda T
MA1-12781 was used in immunohistochemistry to report on a case of primary signet-ring cell carcinoma of the cervix with endometrial and myometrial involvement
Wed Jan 11 00:00:00 EST 2012
Proteomic analysis of zebrafish caudal fin regeneration.
Molecular & cellular proteomics : MCP
Saxena S,Singh SK,Lakshmi MG,Meghah V,Bhatti B,Swamy CV,Sundaram CS,Idris MM
MA1-12781 was used in western blot to study the time-course of zebrafish caudal fin regeneration at the proteomic level
Fri Jun 01 00:00:00 EDT 2012
The value of CDX2 and cytokeratins 7 and 20 expression in differentiating colorectal adenocarcinomas from extraintestinal gastrointestinal adenocarcinomas: cytokeratin 7-/20+ phenotype is more specific than CDX2 antibody.
Diagnostic pathology
Bayrak R,Haltas H,Yenidunya S
MA1-12781 was used in immunohistochemistry to study the value of CDX2 and cytokeratin 7 and 20 expression in differentiating colorectal adenocarcinomas from extraintestinal gastrointestinal adenocarcinomas
Mon Jan 23 00:00:00 EST 2012
Subclassification of non-small cell lung carcinomas lacking morphologic differentiation on biopsy specimens: Utility of an immunohistochemical panel containing TTF-1, napsin A, p63, and CK5/6.
The American journal of surgical pathology
Mukhopadhyay S,Katzenstein AL
MA1-12781 was used in immunohistochemistry to study the utility of an immunohistochemical panel in subtyping non-small cell lung carcinomas lacking morphologic differentiation
Sat Jan 01 00:00:00 EST 2011
Two novel mutations in African and Asian children with progressive familial intrahepatic cholestasis type 3.
Digestive and liver disease : official journal of the Italian Society of Gastroenterology and the Italian Association for the Study of the Liver
Giovannoni I,Santorelli FM,Candusso M,Di Rocco M,Bellacchio E,Callea F,Francalanci P
MA1-12781 was used in immunohistochemistry to investigate the mutations of ABCB4 in progressive familial intrahepatic cholestasis type 3 patients
Fri Jul 01 00:00:00 EDT 2011
Expression and distribution of nucleoside transporter proteins in the human syncytiotrophoblast.
Molecular pharmacology
Errasti-Murugarren E,Díaz P,Godoy V,Riquelme G,Pastor-Anglada M
MA1-12781 was used in immunohistochemistry to determine the localization and function of nucleoside transporter proteins in human syncytiotrophoblast from term placenta.
Tue Nov 01 00:00:00 EDT 2011
The art of identification of extravillous trophoblast.
Placenta
Moser G,Orendi K,Gauster M,Siwetz M,Helige C,Huppertz B
MA1-12781 was used in immunohistochemistry to study the difficulties in identification of extravillous trophoblast
Tue Feb 01 00:00:00 EST 2011
Mucinous tubular and spindle cell carcinoma of kidney and problems in diagnosis.
Turk patoloji dergisi
Sarsik B,Sımşır A,Karaarslan S,Sen S
MA1-12781 was used in immunohistochemistry to study problems in diagnosing mucinous tubular and spindle cell carcinoma of kidney
Sun May 01 00:00:00 EDT 2011
In vitro constitution and in vivo implantation of engineered skin constructs with sweat glands.
Biomaterials
Huang S,Xu Y,Wu C,Sha D,Fu X
MA1-12781 was used in immunohistochemistry to investigate the integration of sweat glands into engineered skin constructs in vitro and in vivo in order to improve the quality of tissue regeneration
Thu Jul 01 00:00:00 EDT 2010
Lung surfactant proteins in the early human placenta.
Histochemistry and cell biology
Sati L,Seval-Celik Y,Demir R
MA1-12781 was used in immunohistochemistry to investigate the distribution of lung surfactant proteins in early human placenta
Fri Jan 01 00:00:00 EST 2010
Fusion of villous trophoblast can be visualized by localizing active caspase 8.
Placenta
Gauster M,Siwetz M,Huppertz B
MA1-12781 was used in immunocytochemistry to examine the change of caspase 8 during the fusion of villous trophoblast
Mon Jun 01 00:00:00 EDT 2009
Antigen expression of human eccrine sweat glands.
Journal of cutaneous pathology
Li HH,Zhou G,Fu XB,Zhang L
MA1-12781 was used in immunohistochemistry to identify the phenotypic markers of human eccrine sweat glands
Sun Mar 01 00:00:00 EST 2009
Giant cell tumor-like lesion of the urinary bladder: a report of two cases and literature review; giant cell tumor or undifferentiated carcinoma?
Diagnostic pathology
Behzatoğlu K,Durak H,Canberk S,Aydin O,Huq GE,Oznur M,Ozyalvaçli G,Yildiz P
MA1-12781 was used in immunohistochemistry to report on two cases of giant cell tumor-like lesion of the urinary bladder
Thu Dec 31 00:00:00 EST 2009
Chondromatous differentiation in clear cell carcinoma of the ovary: a rare finding challenging the differential diagnosis.
Archives of gynecology and obstetrics
Akbulut M,Zekioglu O,Terek MC,Ozdemir N
MA1-12781 was used in immunohistochemistry to investigate a rare case of chondromatous differentiation in an ovarian clear cell carcinoma
Mon Dec 01 00:00:00 EST 2008
Arrangement of collagen fibers in human placental stem villi.
Acta histochemica
Sati L,Demir AY,Sarikcioglu L,Demir R
MA1-12781 was used in immunohistochemistry to investigate the organization of collagen fiber in human placental stem villi
Fri Oct 17 00:00:00 EDT 2008
Mucoepidermoid carcinoma of the parotid gland with ovarian and peritoneal metastasis.
The journal of obstetrics and gynaecology research
Buyukkurt S,Altintas A,Gumurdulu D,Zeren H,Guzel B
MA1-12781 was used in immunohistochemistry to investigate the metastasis of mucoepidermoid carcinoma of the parotid gland
Tue Apr 01 00:00:00 EDT 2008
Isolation, characterization, and differentiation to hepatocyte-like cells of nonparenchymal epithelial cells from adult human liver.
Stem cells (Dayton, Ohio)
Duret C,Gerbal-Chaloin S,Ramos J,Fabre JM,Jacquet E,Navarro F,Blanc P,Sa-Cunha A,Maurel P,Daujat-Chavanieu M
MA1-12781 was used in immunocytochemistry to characterize nonparenchymal epithelial cells from adult human liver
Sun Jul 01 00:00:00 EDT 2007
Combination chemotherapy with gemcitabine and cisplatin as first-line treatment for immunohistochemically proven cholangiocarcinoma.
American journal of clinical oncology
Lee GW,Kang JH,Kim HG,Lee JS,Lee JS,Jang JS
MA1-12781 was used in immunohistochemistry to investigate the combination chemotherapy of gemcitabine and cisplatin against cholangiocarcinoma
Sat Apr 01 00:00:00 EST 2006
Mesenchymal hamartoma of the liver in adulthood: immunohistochemical profiles, clinical and histopathological features in two patients.
Journal of hepato-biliary-pancreatic surgery
Yesim G,Gupse T,Zafer U,Ahmet A
MA1-12781 was used in immunohistochemistry to study two adult patients with mesenchymal hamartoma of the liver
Thu Jul 06 00:00:00 EDT 2006
Isolated idiopathic bile ductular hyperplasia in patients with persistently abnormal liver function tests.
Journal of hepatology
Sonzogni A,Colloredo G,Fabris L,Cadamuro M,Paris B,Roffi L,Pozzi M,Bovo G,Del Poggio P,Portmann BC,Strazzabosco M
MA1-12781 was used in immunohistochemistry to characterize idiopathic isolated ductular hyperplasia in patients undergoing liver biopsy
Thu Apr 01 00:00:00 EST 2004
Protein expression profiles in adenocarcinomas and squamous cell carcinomas of the lung generated using tissue microarrays.
The Journal of pathology
Ullmann R,Morbini P,Halbwedl I,Bongiovanni M,Gogg-Kammerer M,Papotti M,Gabor S,Renner H,Popper HH
MA1-12781 was used in immunohistochemistry to establish the protein expression profiles for lung neoplasms
Thu Jul 01 00:00:00 EDT 2004
CDX-2 homeobox gene expression is a reliable marker of colorectal adenocarcinoma metastases to the lungs.
The American journal of surgical pathology
Barbareschi M,Murer B,Colby TV,Chilosi M,Macri E,Loda M,Doglioni C
MA1-12781 was used in immunohistochemistry to investigate the diagnostic value of CDX-2 in lung metastases from colorectal carcinomas
Sat Feb 01 00:00:00 EST 2003