Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Cytokeratin Pan Type I/II Antibodies

          Progen

          Cytokeratin Pan Type I/II Monoclonal Antibody (Lu-5)

          View all (77) Cytokeratin Pan Type I/II antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Cytokeratin Pan Type I/II Monoclonal Antibody (Lu-5)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (4)
          Cytokeratin Pan Type I/II Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          Cytokeratin Pan Type I/II Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Cytokeratin Pan Type I/II Antibody (610145PROGEN) in IHC

          Immunohistochemistry of mouse tongue (courtesy of J.H, University Hospital Heidelberg). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Cytokeratin Pan Type I/II Antibody in Immunohistochemistry (IHC)
          Cytokeratin Pan Type I/II Antibody in Immunohistochemistry (IHC)
          Cytokeratin Pan Type I/II Antibody in Immunohistochemistry (IHC)
          Cytokeratin Pan Type I/II Antibody in Immunohistochemistry (IHC)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          610145PROGEN

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          Assay-dependent
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:10-1:20
          -

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Bovine, Chicken, Human, Mouse, Reptile, Rat, Xenopus

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          Lu-5

          Immunogen

          Cells Of Lung Cancer Cell Line A549 And A2182

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.05 mg/mL once reconstituted

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 0.5% BSA

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitute lyophilized product with 1 mL distilled water.

          Lu-5 represents an excellent marker for all types of epithelia. Ideal marker for immunolocalization studies on frozen and paraffin embedded tissue. Useful for distinguishing carcinomas from non-epithelial tumors. Tumors specifically detected: all epithelium-derived neoplasms. Polypeptide reacting: Most keratins present in human epithelia and most acidic and basic hair keratins from human hair follicle. The antibody specifically detects a conformational epitope common to most keratins, but not to other intermediate filament polypeptides. Reactivity on cultured cell lines: MCF-7, PLC, A-431, HeLa; PtK2, A6 (XLKE).

          Target Information

          Cytokeratin pan is part of a subfamily of intermediate filament proteins that are characterized by remarkable biochemical diversity, and represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight between 40 kDa- 68 kDa, and an isoelectric pH between 4.9-7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. The composition of cytokeratin pairs vary with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 40-kDa keratin intermediate filament; 47 kDa cytokeratin; 56 kDa cytokeratin; 57kd keratin; 57kDa keratin; 58 kDa cytokeratin; 60-kDa keratin; 63kDa Keratin; 65 kDa cytokeratin; 67 kDa cytokeratin; 70 kd type II; adult keratin; alpha keratin; basic epidermal type II cytokeratin (carboxy-terminal region, clone pUF164); cell proliferation-inducing gene 46 protein; CK-1; CK-10; CK-13; CK-14; CK-15; CK-17; CK-18; CK-19; CK-1B; CK-2e; CK-2P; CK-3; CK-4; CK-5; CK-6C; CK-6E; CK-7; CK-8; cytokeratin 1; cytokeratin 10; cytokeratin 13; cytokeratin 14; cytokeratin 15; cytokeratin 16; cytokeratin 18; cytokeratin 19; cytokeratin 2; cytokeratin 3; cytokeratin 4; cytokeratin 5; cytokeratin 6A; cytokeratin 6C; cytokeratin 6D; Cytokeratin 7; cytokeratin 8; cytokeratin endo A; cytokeratin endo B; cytokeratin otokeratin; cytokeratin type II, component Ia; cytokeratin type II, component Ib/c; cytokeratin type II, component III; cytokeratin VIB; cytokeratin VII; cytokeratin-1; cytokeratin-10; cytokeratin-13; cytokeratin-14; cytokeratin-15; cytokeratin-16; cytokeratin-17; cytokeratin-18; cytokeratin-19; cytokeratin-1B; cytokeratin-2e; cytokeratin-2P; cytokeratin-4; cytokeratin-5; cytokeratin-6A; cytokeratin-6B; cytokeratin-6C; cytokeratin-6E; cytokeratin-7; cytokeratin-8; cytokeratin-A; cytoplasmic intermediate filament protein; cytoskeletal 57 kDa keratin; cytoskeletal protein; ear specific cytokeratin; embryonic type II keratin 1; EndoA cytokeratin (5' end put.); putative; EndoA' cytokeratin (5' end put.); putative; epidermal keratin 10; epidermal keratin type I; epidermal keratin VII; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; epidermolytic hyperkeratosis 1; epidermolytic hyperkeratosis; keratosis palmaris et plantaris; epithelial keratin-2e; fin and gill keratin; focal non-epidermolytic palmoplantar keratoderma; hair alpha protein; HLA-DRB1-restricted epitope S1; HLA-DRB1-restricted epitope S2; HLA-DRB1-restricted epitope S3; HLA-DRB1-restricted epitope S4; HMWCK; intermediate filament; intermediate filament protein; intermediate filament protein K16; intermediate filament protein; K7; K1; K10; K13; K14; K15; K17; K18; K19; K1B; K2e; K2P; K5; K6-alpha; K6a keratin; K6C; K7; K76; K77; K8; KERA; keratin 1, type II; keratin 10 (epidermolytic hyperkeratosis); keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris); keratin 10, type I; keratin 10, type I L homeolog; keratin 10, type I S homeolog; keratin 13, type I; keratin 13, type I S homeolog; keratin 14; keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner); keratin 14, type I; keratin 14, type I L homeolog; keratin 14-like; keratin 15; keratin 15, type I; keratin 16 (focal non-epidermolytic palmoplantar keratoderma); keratin 16 homolog; keratin 16, type I; keratin 16, type I S homeolog; keratin 17, type I; keratin 17, type I L homeolog; keratin 18, type I; keratin 19; keratin 19, type I; Keratin 1B; keratin 2 epidermis; keratin 2, type II; keratin 2p; keratin 3, type II; keratin 4, type II; keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5 L homeolog; keratin 5, gene 2; keratin 5, type II; keratin 6 alpha; keratin 6 L homeolog; keratin 6A, , type II; keratin 6A, type II; keratin 6C, type II; keratin 6E; keratin 7, type II; keratin 76, type II; keratin 77, type II; keratin 8; keratin 8, type II; keratin 86; keratin complex 1, acidic, gene 10; keratin complex 1, acidic, gene 13; keratin complex 1, acidic, gene 14; keratin complex 1, acidic, gene 15; keratin complex 1, acidic, gene 16; keratin complex 1, acidic, gene 17; keratin complex 1, acidic, gene 18; keratin complex 1, acidic, gene 19; keratin complex 2, basic, gene 1; keratin complex 2, basic, gene 17; keratin complex 2, basic, gene 39; keratin complex 2, basic, gene 4; keratin complex 2, basic, gene 5; keratin complex 2, basic, gene 6a; keratin complex 2, basic, gene 6c; keratin complex 2, basic, gene 7; keratin complex 2, basic, gene 8; keratin complex 2, gene 17; keratin D; keratin intermediate filament 16a; keratin intermediate filament 16b; keratin IV; keratin K5; keratin K6; keratin K6h; keratin L homeolog; keratin S homeolog; keratin type II; Keratin type II cytoskeletal 5; keratin, 55K type II cytoskeletal; keratin, epidermal type II, K6A; keratin, simple epithelial type I, K7; keratin, type I cytoskeletal 59 kDa; keratin, type I, 40-kd; keratin, type II cytoskeletal 5; keratin, type II cytoskeletal 5-like; keratin, type II cytoskeletal 60 kDa, component III; keratin, type II cytoskeletal 6A; keratin-10; keratin-15, basic; keratin-15, beta; keratin-2 epidermis; keratin-2e; keratin-6-alpha; KRT1B; KRT5; mK6-alpha; ORF; otokeratin; ouroboros 2; Pan CK; pan Cytokeratin; pan keratin; pankeratin; PIG46; Sak57; sarcolectin; suprabasal cytokeratin 10; tuftelin interacting protein 8; type I alpha-keratin 14; type I alpha-keratin 15; type I cytoskeletal 15; type I keratin KA10; type I keratin KA13; type I keratin Ka14; type I keratin KA15; type I keratin KA16; type I keratin KA17; type I keratin KA19; type II alpha-keratin IIA; type II alpha-keratin IIC; type II cytokeratin A (no.8); type II cytoskeletal 1b; type II cytoskeletal cochleal; type II keratin Kb2; type II keratin Kb39; type II keratin Kb9; type II keratin subunit protein; type II mesothelial keratin K7; type-II intermediate filament protein; type-II keratin Kb1; type-II keratin Kb12; type-II keratin Kb2; type-II keratin Kb3; Type-II Keratin Kb39; type-II keratin Kb4; type-II keratin Kb5; type-II keratin Kb6; type-II keratin Kb7; type-II keratin Kb8; type-II keratin Kb9; unnamed protein product

          View more View less

          Gene Aliases: 2310001L23Rik; 2310016L08Rik; 3300001P10Rik; 39.1; 4732484G22Rik; AEI2; BCIE; BIE; CARD2; CK 2e; CK-14; CK-16; CK-17; CK-18; CK-19; CK-1B; CK-2e; CK-4; CK-5; CK-6A; CK-6C; CK-6E; CK-8; CK1; CK10; CK13; CK14; CK15; CK16; CK18; CK19; CK3; CK4; CK5; CK6A; CK6C; CK6D; CK7; CK8; CYK18; CYK4; CYK8; CYKER; D130054E02Rik; D15Wsu77e; DDD; DDD1; EBS1; EBS1A; EBS1B; EBS1C; EBS1D; EBS2; EBS2A; EBS2B; EBS2C; EBS2D; EBS2E; EBS2F; EBS3; EBS4; EHK; EHK1; EHK2; EHK2A; EHK2B; EndoA; EndoC; EPPK; fgk; FNEPPK; GK-19; HUMCYT2A; IHL; K1; K10; K13; K14; K15; K16; K17; K18; K19; K1B; K1C1; K1CO; K1CP; K1CS; K1emb; K2C7; K2C8; K2e; K3; K4; K5; K6[a]; K6A; K6C; K6D; K6E; K7; K76; K77; K8; Ka10; Ka13; Ka14; Ka15; Ka16; Ka17; Ka19; Kb1; Kb2; Kb39; Kb4; Kb9; keratin; keratin-19; KERIA; KERIB; KO; KPP; krt; Krt-1.10; Krt-1.13; Krt-1.14; Krt-1.19; Krt-2.1; Krt-2.4; Krt-2.8; krt-a; krt-b; krt.L; krt.S; Krt1-10; Krt1-13; Krt1-14; Krt1-15; Krt1-16; Krt1-17; Krt1-18; Krt1-19; krt1-2; krt10; krt10.L; krt10.S; krt13; krt13.S; Krt14; krt14.L; Krt14l; KRT15; krt16; krt16.S; KRT16A; krt17; KRT19; KRT1A; KRT1B; Krt2-1; Krt2-17; Krt2-2; Krt2-4; Krt2-5; Krt2-6a; Krt2-6c; Krt2-7; KRT2-8; Krt239; KRT2A; KRT2B; KRT2E; KRT2P; krt5.2; krt5.2.L; KRT5A; krt6; krt6.L; Krt6a; KRT6C; KRT6D; KRT6E; Krt86; krt9; KRTE; MECD2; mK1b; mK6[a]; MK6a; NEPPK; NFJ; ouro2; PC1; PC2; PC3; PCHC1; PPKNEFD; pUF164; Sak57; SCL; Tfip8; TROMA-1; WSN1; WSN2; XELAEV_18013614mg; XELAEV_18031303mg; XELAEV_18043105mg; XELAEV_18043108mg; XELAEV_18044427mg; XELAEV_18045881mg; XELAEV_18045882mg; XELAEV_18045886mg; XELAEV_18045894mg; xk70

          View more View less

          UniProt ID: (Human) P04264, (Bovine) P06394, (Human) P13645, (Human) P13646, (Human) P02533, (Chicken) Q6PVZ1, (Human) P19012, (Human) P08779, (Human) Q04695, (Bovine) A1L595, (Human) P05783, (Bovine) P08728, (Human) P08727, (Human) P35908, (Chicken) O93256, (Human) P12035, (Human) P19013, (Bovine) Q5XQN5, (Human) P13647, (Human) P02538, (Human) P48668, (Human) P08729, (Chicken) O93532, (Bovine) Q29S21, (Human) Q01546, (Human) Q7Z794, (Bovine) P05786, (Human) P05787, (Rat) Q6IMF3, (Mouse) P04104, (Mouse) P02535, (Rat) Q6IFW6, (Mouse) P08730, (Rat) Q6IFV4, (Mouse) Q61781, (Rat) Q6IFV1, (Mouse) Q61414, (Rat) Q6IFV3, (Mouse) Q9Z2K1, (Mouse) Q9QWL7, (Rat) Q6IFU8, (Mouse) P05784, (Rat) Q5BJY9, (Mouse) P19001, (Rat) Q63279, (Rat) Q6IG02, (Mouse) Q3TTY5, (Rat) Q6IG00, (Mouse) P07744, (Rat) Q6P6Q2, (Mouse) Q922U2, (Mouse) P50446, (Rat) Q4FZU2, (Rat) Q6IG12, (Mouse) Q9DCV7, (Mouse) Q3UV17, (Rat) Q6IG01, (Mouse) Q6IFZ6, (Rat) Q10758, (Mouse) P11679

          View more View less

          Entrez Gene ID: (Human) 3848, (Bovine) 281888, (Chicken) 771977, (Human) 3858, (Human) 3860, (Chicken) 408040, (Human) 3861, (Bovine) 404111, (Chicken) 408039, (Bovine) 507464, (Human) 3866, (Human) 3868, (Human) 3872, (Chicken) 100858439, (Bovine) 281889, (Bovine) 506480, (Human) 3875, (Chicken) 101749333, (Bovine) 514812, (Human) 3880, (Human) 3849, (Chicken) 395861, (Bovine) 407188, (Human) 3850, (Bovine) 527819, (Human) 3851, (Bovine) 281268, (Chicken) 407779, (Human) 3852, (Bovine) 614456, (Human) 3853, (Chicken) 408041, (Human) 286887, (Human) 3855, (Chicken) 395772, (Bovine) 535697, (Bovine) 407189, (Human) 51350, (Human) 374454, (Bovine) 281269, (Chicken) 426896, (Human) 3856, (Rat) 300250, (Mouse) 16678, (Mouse) 16661, (Rat) 450225, (Mouse) 16663, (Rat) 287699, (Mouse) 16664, (Rat) 287701, (Mouse) 16665, (Rat) 287700, (Rat) 303530, (Mouse) 16666, (Mouse) 16667, (Rat) 287702, (Mouse) 16668, (Rat) 294853, (Mouse) 16669, (Rat) 360626, (Rat) 406228, (Mouse) 16681, (Rat) 315323, (Mouse) 16682, (Rat) 369017, (Mouse) 110308, (Rat) 120093743, (Mouse) 16687, (Rat) 683313, (Rat) 300242, (Mouse) 110310, (Rat) 407757, (Mouse) 77055, (Rat) 406226, (Mouse) 406220, (Rat) 25626, (Mouse) 16691, (Xenopus) 446743, (Xenopus) 444694, (Xenopus) 495266, (Xenopus) 734797, (Xenopus) 779233, (Xenopus) 100381074, (Xenopus) 495267, (Xenopus) 380402, (Xenopus) 397907, (Xenopus) 100310818

          View more View less

          Function(s)
          protein binding carbohydrate binding structural constituent of epidermis signaling receptor activity protein heterodimerization activity structural molecule activity structural constituent of cytoskeleton keratin filament binding scaffold protein binding RNA binding structural constituent of muscle cadherin binding involved in cell-cell adhesion macromolecular complex binding cytoskeletal protein binding
          Process(es)
          keratinization intermediate filament cytoskeleton organization intermediate filament organization morphogenesis of an epithelium cytoskeleton organization epithelial cell differentiation regulation of translation in response to stress Notch signaling pathway response to estrogen sarcomere organization cell differentiation involved in embryonic placenta development wound healing negative regulation of keratinocyte differentiation biological_process keratinocyte development epidermis development keratinocyte differentiation positive regulation of epidermis development protein heterotetramerization cornification cellular response to calcium ion tumor necrosis factor-mediated signaling pathway response to hydrostatic pressure response to other organism epithelium development extrinsic apoptotic signaling pathway hepatocyte apoptotic process negative regulation of hepatocyte apoptotic process tongue morphogenesis cellular response to retinoic acid response to mechanical stimulus regulation of cell migration regulation of protein localization intermediate filament polymerization complement activation, lectin pathway negative regulation of inflammatory response establishment of skin barrier hair cycle intermediate filament bundle assembly positive regulation of keratinocyte differentiation positive regulation of cell growth hair follicle morphogenesis positive regulation of translation positive regulation of hair follicle development response to radiation negative regulation of epithelial cell proliferation positive regulation of cell proliferation cell differentiation negative regulation of cell migration killing of cells of other organism defense response to Gram-positive bacterium antimicrobial humoral immune response mediated by antimicrobial peptide negative regulation of entry of bacterium into host cell Wnt signaling pathway Golgi to plasma membrane protein transport negative regulation of apoptotic process anatomical structure morphogenesis cell-cell adhesion pigmentation sebaceous gland development inflammatory response innate immune response keratinocyte migration keratinocyte activation keratinocyte proliferation intermediate filament-based process response to fructose response to nutrient levels hepatocyte differentiation mesenchymal stem cell differentiation cholangiocyte apoptotic process response to zinc ion response to ionizing radiation negative regulation of epithelial cell differentiation response to oxidative stress fibrinolysis regulation of angiogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-5d76f9f486-n5jxz:80/100.66.73.31:80.
          git-commit: 08b979eaab39376f31a39358ffa32de998e3c7f7
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.6.0-offline-prod