Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4ºC in the dark. Once reconstituted, this product has a shelf life of approx. 18 months.
For FACS analysis, use 10 µL of the suggested working dilution to label 1x10^6 cells in 100 µL.
Rat anti Mouse Dectin-1 antibody, clone 2A11 recognizes murine beta-glucan receptor, also known as Dectin-1.
Rat anti Mouse Dectin-1 antibody, clone 2A11 recognizes murine beta-glucan receptor, also known as Dectin-1.
CLEC7A, also known as dectin-1, is a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily and is predominantly expressed on myeloid cells. It is a small glycoprotein type II membrane receptor with an extracellular C-type lectin-like domain fold and a cytoplasmic domain with an immunoreceptor tyrosine-based activation motif (ITAM). CLEC7A functions as a pattern-recognition receptor that recognizes a variety of beta-1,3-linked and beta-1,6-linked glucans from fungi and plants, and in this way plays a role in innate immune response. Upon fungal exposure, CLEC7A activates Syk tyrosine kinase, triggering a massive oxidative burst through the formation of reactive oxygen species.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: beta-glucan receptor; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 12; C-type lectin domain family 7 member A; C-type lectin superfamily member 12; DC-associated C-type lectin 1; dectin-1; dendritic cell-associated C-type lectin 1
Gene Aliases: beta-GR; BGR; Clecsf12
Entrez Gene ID: (Mouse) 56644
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support