Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: GDERFYAEHL MPTLQGLLDP ESAHRLAVRF TSLGLLPRAR FQDSDMLEVR VLGHKFRNPV GIAAGFDKHG EAVDGLYKMG FGFVEIGSVT PKPQEGNPRP RVFRLPEDQ
The protein encoded by this gene catalyzes the fourth enzymatic step, the ubiquinone-mediated oxidation of dihydroorotate to orotate, in de novo pyrimidine biosynthesis. This protein is a mitochondrial protein located on the outer surface of the inner mitochondrial membrane. [provided by RefSeq, Jul 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DHOdehase; Dihydroorotate dehydrogenase (quinone), mitochondrial; Dihydroorotate oxidase; human complement of yeast URA1
Gene Aliases: DHOdehase; DHODH; POADS; URA1
UniProt ID: (Human) Q02127
Entrez Gene ID: (Human) 1723
Molecular Function: dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support