Search
Search
Bethyl Laboratories
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
HELB gene ontology annotations related to this gene include cellular response to DNA damage stimulus; DNA replication, synthesis of RNA primer; negative regulation of double-strand break repair via homologous recombination; regulation of DNA double-strand break processing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA helicase B; hDHB; helicase (DNA) B
Gene Aliases: AI447783; D10Ertd664e; DHB; hDHB; HELB
UniProt ID: (Human) Q8NG08, (Mouse) Q6NVF4
Entrez Gene ID: (Human) 92797, (Mouse) 117599
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support