Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For maximum recovery of product, centrifuge the vial prior to removing the cap.
Sequence of this protein is as follows: GPRMSQNLLR QADLDKF
This gene encodes an inner dynein arm heavy chain that provides structural support between the radial spokes and the outer doublet of the sperm tail. Naturally occurring mutations in this gene are associated with primary ciliary dyskinesia and multiple morphological anomalies of the flagella that result in asthenozoospermia and male infertility. Mice with a homozygous knockout of the orthologous gene are viable but have reduced sperm motility and are infertile.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Axonemal beta dynein heavy chain 1; Ciliary dynein heavy chain 1; DNAH1 variant protein; Dynein axonemal heavy chain 1; dynein, axonemal, heavy chain 1; dynein, axonemal, heavy polypeptide 1; hDHC7; Heat shock regulated protein 1; HSRF-1; testicular tissue protein Li 60
Gene Aliases: DHC7; DNAH1; DNAHC1; HDHC7; HL-11; HL11; HSRF-1; KIAA1410; XLHSRF-1
UniProt ID: (Human) Q9P2D7
Entrez Gene ID: (Human) 25981
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support