Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For maximum recovery of product, centrifuge the vial prior to removing the cap.
Sequence of this protein is as follows: PVSVEFIRKH TKELSPTSDT
Force generating protein of respiratory cilia. Produces force towards the minus ends of microtubules. Dynein has ATPase activity; the force-producing power stroke is thought to occur on release of ADP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Axonemal beta dynein heavy chain 7; axonemal dynein heavy chain 7; Ciliary dynein heavy chain 7; Dynein axonemal heavy chain 7; Dynein heavy chain-like protein 2; dynein, axonemal, heavy chain 7; dynein, axonemal, heavy polypeptide 7; hDHC2
Gene Aliases: DNAH7; KIAA0944
UniProt ID: (Human) Q8WXX0
Entrez Gene ID: (Human) 56171
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support