Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Dynamin 2 Antibodies

          Proteintech

          DNM2 Monoclonal Antibody (1B8H3), CoraLite® Plus 647

          View all (21) Dynamin 2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite DNM2 Monoclonal Antibody (1B8H3), CoraLite® Plus 647

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          DNM2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          DNM2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DNM2 Antibody (CL647-68209) in ICC/IF

          Immunofluorescent analysis of (-20°C Methanol) fixed HeLa cells using CoraLite® Plus 647 DNM2 antibody (CL647-68209, Clone: 1B8H3) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DNM2 Antibody in Immunocytochemistry (ICC/IF)
          DNM2 Monoclonal Antibody (1B8H3), CoraLite® Plus 647

          Product Details

          CL647-68209

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Pig, Rabbit, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1B8H3

          Immunogen

          DNM2 fusion protein Ag6274
          3D Epitope / Immunogen

          Conjugate

          CoraLite® Plus 647 CoraLite® Plus 647 CoraLite® Plus 647
          • Unconjugated
          • CoraLite 555
          • CoraLite 594
          • Request custom conjugation

          Excitation/Emission Max

          654/674 nm

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          PBS, pH 7.3, with 50% glycerol, 0.5% BSA

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, store in dark

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: RRGWLTINNI SLMKGGSKEY WFVLTAESLS WYKDEEEKEK KYMLPLDNLK IRDVEKGFMS NKHVFAIFNT EQRNVYKDLR QIELACDSQE DVDSWKASFL RAGVYPEKDQ AENEDGAQEN TFSMDPQLER QVETIRNLVD SYVAIINKSI RDLMPKTIMH LMINNTKAFI HHELLAYLYS SADQSSLMEE SADQAQRRDD MLRMYHALKE ALNIIGDIST STVSTPVPPP VDDTWLQSAS SHSPTPQRRP VSSIHPPGRP PAVRGPTPGP PLIPVPVGAA ASFSAPPIPS RPGPQSVFAN SDLFPAPPQI PSRPVRIPPG IPPGVPSRRP PAAPSRPTII RPAEPSLLD

          Target Information

          Dynamins represent one of the subfamilies of GTP-binding proteins. These proteins share considerable sequence similarity over the N-terminal portion of the molecule, which contains the GTPase domain. Dynamins are associated with microtubules. They have been implicated in cell processes such as endocytosis and cell motility, and in alterations of the membrane that accompany certain activities such as bone resorption by osteoclasts. Dynamins bind many proteins that bind actin and other cytoskeletal proteins. Dynamins can also self-assemble, a process that stimulates GTPase activity. Five alternatively spliced transcripts encoding different proteins have been described. Additional alternatively spliced transcripts may exist, but their full-length nature has not been determined.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Dynamin 2; Dynamin II; Dynamin UDNM; Dynamin-2; unnamed protein product

          View more View less

          Gene Aliases: b2b2159Clo; CMT2M; CMTDI1; CMTDIB; DI-CMTB; DNM2; DYIIAAB; DYN2; DYNII; LCCS5; Udnm

          View more View less

          UniProt ID: (Human) P50570, (Rat) P39052, (Mouse) P39054

          View more View less

          Entrez Gene ID: (Rabbit) 100348451, (Human) 1785, (Pig) 100622538, (Rat) 25751, (Mouse) 13430

          View more View less

          Function(s)
          nucleotide binding GTPase activity protein binding GTP binding phosphatidylinositol-4,5-bisphosphate binding microtubule binding hydrolase activity SH3 domain binding enzyme binding protein kinase binding D2 dopamine receptor binding protein complex binding phosphatidylinositol 3-kinase regulatory subunit binding WW domain binding nitric-oxide synthase binding membrane traffic protein
          Process(es)
          G2/M transition of mitotic cell cycle regulation of transcription, DNA-templated post-Golgi vesicle-mediated transport endocytosis receptor-mediated endocytosis phagocytosis autophagy centrosome cycle signal transduction synaptic vesicle budding from presynaptic endocytic zone membrane antigen processing and presentation of exogenous peptide antigen via MHC class II regulation of axon extension receptor internalization transferrin transport positive regulation of apoptotic process stress fiber assembly positive regulation of transcription, DNA-templated synaptic vesicle transport neuron projection morphogenesis protein polymerization membrane organization actin filament bundle organization membrane tubulation vesicle scission negative regulation of membrane tubulation G-protein coupled receptor internalization Golgi to plasma membrane transport spermatogenesis metabolic process response to light stimulus positive regulation of lamellipodium assembly negative regulation of transforming growth factor beta receptor signaling pathway regulation of Rac protein signal transduction response to cocaine macropinocytosis positive regulation of nitric oxide biosynthetic process positive regulation of endocytosis positive regulation of phagocytosis cilium morphogenesis cellular response to carbon monoxide cellular response to X-ray cellular response to nitric oxide positive regulation of substrate adhesion-dependent cell spreading cellular response to dopamine regulation of Golgi organization positive regulation of sodium:potassium-exchanging ATPase activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline