Applications | Tested Dilution | Publications |
---|---|---|
Immunohistochemistry (IHC) |
Assay-dependent | View 2 publications 2 publications |
ELISA (ELISA) |
Assay-dependent | - |
Product Specifications | |
---|---|
Species Reactivity |
Chemical |
Published species |
Chemical |
Host/Isotype |
Rat / IgE |
Class |
Monoclonal |
Type |
Antibody |
Clone |
LO-DNP-30 |
Immunogen |
Synthetic DNP |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
purified |
Storage buffer |
PBS, pH 7.4 |
Contains |
0.1% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_2532965 |
DNP (2,4-dinitrophenol) is a toxic dye, chemically related to trinitrophenol (picric acid). It is used in biochemical studies of oxidative processes where it uncouples oxidative phosphorylation and is also used as a metabolic stimulant. 2,4-Dinitrophenol is used in the manufacture of dyes, wood preservatives, and as a pesticide. The acute (short-term) effects of 2,4-dinitrophenol in humans through oral exposure are nausea, vomiting, sweating, dizziness, headaches, and loss of weight. Chronic (long-term) oral exposure to 2,4-dinitrophenol in humans has resulted in the formation of cataracts and skin lesions, weight loss, and has caused effects on the bone marrow, central nervous system (CNS), and cardiovascular system. [National Center for Biotechnology Information. PubChem Compound Database; CID=1493].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C6H3N2O4; C6H4N2O5; Dinitrophenol; DNP
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support