Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MLANSASVRI LIKGDKVVND DCTHEADVYI ENGIIQQVGR ELMIPGGAKV IDATGKLVIP GGIDTSTHFH QTFMNATCVD DFYHGTKAAL VGGTTMIIGH VLPDKETSLV DAYEKCRGLA DPKVCCDYAL HVGITWWAPK VKAEMETLVR EKGVNSFQMF MTYKDLYMLR DSELYQVLHA CKDIGAIARV HAENGELVAE GAKEALDLGI TGPEGIEISR PEELEAEATH RVITIANRTH CPIYLVNVSS ISAGDVIAAA KMQGKVVLAE TTTAHATLTG LHYYHQDWSH AAAYVTVPPL RLDTNTSTYL MSLLANDTLN IVASDHRPFT TKQKAMGKED FTKIPHGVSG VQDRMSVIWE RGVVGGKMDE NRFVAVTSSN AAKLLNLYPR KGRIIPGADA DVVVWDPEAT KTISASTQVQ GGDFNLYENM RCHGVPLVTI SRGRVVYENG VFMCAEGTGK FCPLRSFPDT VYKKLVQREK TLKVRGVDRT PYLGDVAVVV HPGKKEMGTP LADTPTRPVT RHGGMRDLHE SSFSLSGSQI DDHVPERASA RILAPPGGRS SGIW
The collapsin response mediator protein (CRMP) family of proteins (also referred to as ULIP6) are a group of neurologic proteins that have been found to play key roles in growth cone guidance during neural development. CRMP5 has relatively low sequence homology with the other four members of the CRMP family. CRMP5 is expressed in the developing nervous system in an expression pattern that resembles CRMP2 and has a peak expression during the first postnatal week. Elevated levels of autoantibody to CRMP5 have been shown to be linked to paraneoplastic autoimmune optic neuritis and other paraneoplastic neoplasms. Auto-immunity against CRMP5 expression (predominantly the N-terminal epitopes) is also being investigated as an oncological marker in the respiratory system and in the thymus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Collapsin response mediator protein 5; collapsin response mediator protein-5; CRAM; CRMP-5; CRMP3-associated molecule; dihydropyrimidinase-like 5; Dihydropyrimidinase-related protein 5; DRP-5; ULIP-6; ULIP6 protein; UNC33-like phosphoprotein 6
Gene Aliases: CRAM; CRMP-5; CRMP5; DPYSL5; ULIP6
UniProt ID: (Human) Q9BPU6, (Rat) Q9JHU0
Entrez Gene ID: (Human) 56896, (Rat) 65208
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support