Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000 | - |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:50 | - |
Flow Cytometry (Flow) |
1:200 | - |
Immunoprecipitation (IP) |
1:50 | - |
Product Specifications | |
---|---|
Species Reactivity |
Mouse |
Host/Isotype |
Rabbit / IgG |
Class |
Monoclonal |
Type |
Antibody |
Clone |
E.959.6 |
Immunogen |
Synthetic peptide corresponding to residues surrounding |
Conjugate |
Unconjugated |
Form |
Liquid |
Purification |
Affinity chromatography |
Storage buffer |
0.01M HEPES, pH 7.5, with 0.15M NaCl, 100µg/mL BSA, 50% glycerol |
Contains |
<0.02% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_11002186 |
It is not recommended to aliquot this antibody.
Protein kinases play important roles in the signal transduction in response to a variety of external stimuli. Recently, several protein kinases have been identified that may also be involved in apoptotic process. Overexpression of a serine/threonine kinase, ZIP kinase can cause the morphological changes typical of apoptosis in NIH 3T3 cells. Sanjo et al, have identified two new protein kinases DRAK1 and DRAK2 (Death Receptor Associated Kinase 1 and 2). Both DRAKs and ZIP kinase share significant homology at the amino acid level. The kinase domains of both DRAKs, ZIP kinase are homologous to DAP (Death-associated protein) kinase, which is involved in the apoptotic signaling induced by interferon-g. Overexpression of DRAKs in NIH 3T3 cells lead to apoptosis. These transfection experiments suggest that C-terminal domain of DRAKs are important for induction of apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DAP kinase-related apoptosis-inducing protein kinase 2; DRAK 2; Serine/threonine-protein kinase 17B; ST17B; STK 17B
Gene Aliases: 3110009A03Rik; AI120141; Drak2; Stk17b
UniProt ID: (Mouse) Q8BG48
Entrez Gene ID: (Mouse) 98267
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support