Search
Search
Exalpha
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive control: Placenta.
The product of the DEGS1 gene is a member of the membrane fatty acid (sphingolipid delta 4 ) desaturase family. Reported to function by inserting double bonds at delta 4 position in fatty acids. DEGS1 is an intracellular enzyme. Overexpression of this gene inhibited biosynthesis of the EGF receptor, suggesting a possible role of a fatty acid desaturase in regulation biosynthetic processing of the EGF receptor. Two splice variants have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell migration-inducing gene 15 protein; Degenerative spermatocyte homolog 1; degenerative spermatocyte homolog 1, lipid desaturase; degenerative spermatocyte homolog, lipid desaturase; Degenerative spermatocyte-like protein RDES; Delta(4)-desaturase; dihydroceramide desaturase; dihydroceramide desaturase 1; Dihydroceramide desaturase-1; DSH1; membrane fatty acid (lipid) desaturase; Membrane lipid desaturase; MGC5079; MIG15; migration-inducing gene 15 protein; putative fatty acid desaturase; Retinol isomerase; sphingolipid 1; sphingolipid delta 4 desaturase; sphingolipid delta(4)-desaturase 1; Sphingolipid delta(4)-desaturase DES1
Gene Aliases: AA536663; DEGS; DEGS-1; DEGS1; Des-1; DES1; FADS7; HLD18; Mdes; MIG15; MLD
UniProt ID: (Human) O15121, (Mouse) O09005, (Rat) Q91XI6
Entrez Gene ID: (Human) 8560, (Mouse) 13244, (Rat) 58970
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support