Applications | Tested Dilution | Publications |
---|---|---|
Immunohistochemistry (Paraffin) (IHC (P)) |
Assay-dependent | - |
Immunohistochemistry (Frozen) (IHC (F)) |
Assay-dependent | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Mouse / IgG2a |
Class |
Monoclonal |
Type |
Antibody |
Clone |
Dsg1-P124 |
Immunogen |
Recombinant Human Desmoglein 1 (Extracellular Part Eii, Eiii And Eiv) |
Conjugate |
Unconjugated |
Form |
Liquid |
Storage conditions |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Recognizes human skin the, specifically with a 160 kD polypeptide typical for Desmoglein 1. In immunolocalization studies the mab shows typical staining of suprabasal layers of the epidermis and special regions of fetal anagen hair follicles. The staining pattern observed corresponds to those obtained with pemphigus foliaceus patient sera. Cross-reaction with Desmogleins 2 and 3 was not detected.
Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cadherin family member 4; Desmoglein-1; Desmosomal glycoprotein 1; DG1; Pemphigus foliaceus antigen
Gene Aliases: CDHF4; DSG1
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support