Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500 - 1:2,000 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rat |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Recombinant fusion protein containing a sequence corresponding to amino acids 635-935 of human DSG2 (NP_001934.2). |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
4.96 mg/mL |
Purification |
Affinity Chromatography |
Storage buffer |
PBS, pH 7.3, with 50% glycerol |
Contains |
0.01% thimerosal |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
RRID |
AB_2914804 |
Positive test controls include: HepG2, HeLa, SW620, DU145, K-562. The target is usually found in the following locations: Cell junction, Cell membrane, Single-pass type I membrane protein, desmosome.
Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARVC10; ARVD10; Cadherin family member 5; CMD1BB; desmoglein 2; Desmoglein-2; DSG-2; HDGC; MGC117034; MGC117036; MGC117037
Gene Aliases: AA408168; CDHF5; D18Ertd293e; DSG2; HDGC
UniProt ID: (Human) Q14126, (Mouse) O55111
Entrez Gene ID: (Human) 1829, (Mouse) 13511, (Rat) 307562
Molecular Function:
cadherin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support