Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The D1 subtype is the most abundant dopamine receptor in the central nervous system. This G-protein coupled receptor stimulates adenylyl cyclase and activates cyclic AMP-dependent protein kinases. D1 receptors regulate neuronal growth and development, mediate some behavioral responses, and modulate dopamine receptor D2-mediated events. Alternate transcription initiation sites result in two transcript variants of this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D(1B) dopamine receptor; D(5) dopamine receptor; D1b receptor; Dopamine 5 receptor; Dopamine D5 receptor; dopamine receptor 5; DRD 1B; DRD 5; Hd5dr; MGC10601
Gene Aliases: D1B; D5R; Drd-5; Drd1b; Drd5; Gpcr1
UniProt ID: (Mouse) Q8BLD9, (Rat) P25115
Entrez Gene ID: (Mouse) 13492, (Rat) 25195
Molecular Function: G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support