Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000-1:5,000 | - |
Product Specifications | |
---|---|
Species Reactivity |
Bacteria |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Recombinant Escherichia coli O157:H7 Outer membrane protein C protein (22-367aa) |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
4.56 mg/mL |
Purification |
Protein G |
Storage buffer |
10mM PBS, pH 7.4, with 50% glycerol |
Contains |
0.03% ProClin 300 |
Storage conditions |
-20°C or -80°C if preferred |
RRID |
AB_2902330 |
This antibody has been tested in direct-ELISA
Forms passive diffusion pores which allow small molecular weight hydrophilic materials across the outer membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support