Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500-1:1,000 | View 3 publications 3 publications |
Immunohistochemistry (IHC) |
Assay-dependent | View 1 publication 1 publication |
Immunoprecipitation (IP) |
0.5-4.0 µg | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rat |
Published species |
Human |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
E2F3 Fusion Protein Ag3039 (1-110 aa encoded by BC016847) |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
0.13 mg/mL |
Storage conditions |
-20°C |
Immunogen sequence: MQSGGGVKT DDTSTLNSLC GYAWVYVWEE KQRCRLSSFF SSSASIPGLL PSHTLDLVQN VGVVLDEALG WGRERELCVK CLLEMHCGVF SCMGNHLCQA FPHFPYLSHL V (1-110 aa encoded by BC016847)
The protein encoded by this gene is a member of the E2F family of transcription factors. The E2F family plays a crucial role in the control of cell cycle and action of tumor suppressor proteins and is also a target of the transforming proteins of small DNA tumor viruses. The E2F proteins contain several evolutionally conserved domains found in most members of the family. These domains include a DNA binding domain, a dimerization domain which determines interaction with the differentiation regulated transcription factor proteins (DP), a transactivation domain enriched in acidic amino acids, and a tumor suppressor protein association domain which is embedded within the transactivation domain. This protein and another 2 members, E2F1 and E2F2, have an additional cyclin binding domain. This protein binds specifically to retinoblastoma protein pRB in a cell-cycle dependent manner.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686C18211; E2F-3; MGC104598; RP1-177P22.2; Transcription factor E2F3
Gene Aliases: E2F3; KIAA0075
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support