Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
2 µg/ml | View 3 publications 3 publications |
Immunohistochemistry (IHC) |
- | View 2 publications 2 publications |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:20 | - |
Gel Shift (GS) |
1mg/mL | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rat |
Published species |
Human, Rat |
Host/Isotype |
Mouse / IgG1 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
4E2F04 (WUF10) |
Immunogen |
Recombinant human E2F-4 protein |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
0.2 mg/mL |
Purification |
Protein G |
Storage buffer |
PBS, pH 7.4, with 0.2% BSA |
Contains |
0.09% sodium azide |
Storage conditions |
4° C |
RRID |
AB_10984014 |
MA5-11276 targets E2F-4 Transcription Factor in GS, IF, IHC (P), and WB applications and shows reactivity with Human and Rat samples.
The MA5-11276 immunogen is recombinant human E2F-4 protein.
E2F transcription factors are functionally regulated by binding to Rb p110, p107, and p130. E2F-4 is regulated by complex formation with Rb p110. E2F family members bind DNA as heterodimers with members of the DP family of polypeptides Differential phosphorylation contributes to the microheterogeniety of E2F-4 (total aa 411-416, varies due to a trinucleotide repeat).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E2F transcription factor 4, p107/p130-binding; E2F-4; p107/p130-binding protein; Transcription factor E2F4
Gene Aliases: 2010111M04Rik; AI427446; E2F-4; E2F4
UniProt ID: (Human) Q16254, (Mouse) Q8R0K9
Entrez Gene ID: (Human) 1874, (Rat) 100360427, (Mouse) 104394
Molecular Function:
RNA metabolism protein
nucleic acid metabolism protein
general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support