Applications | Tested Dilution | Publications |
---|---|---|
Flow Cytometry (Flow) |
0.25 µg/1x10^6 cells | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Rat / IgG1 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
303913 |
Immunogen |
Mouse myeloma cell line NS0-derived recombinant human ECE-1 Gln90-Trp770 |
Conjugate |
Unconjugated |
Form |
Lyophilized |
Concentration |
0.5 mg/mL |
Purification |
Protein A/G |
Storage buffer |
PBS with 5% trehalose |
Contains |
No Preservative |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
RRID |
AB_2608781 |
Reconstitute at 0.5 mg/mL in sterile PBS.
ECE-1 is involved in proteolytic processing of endothelin precursors to biologically active peptides.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ECE 1; ECE-1; Endothelin-converting enzyme 1
Gene Aliases: ECE; ECE1
UniProt ID: (Human) P42892
Entrez Gene ID: (Human) 1889
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support