Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • EFTUD2 Antibodies

          Invitrogen

          EFTUD2 Recombinant Rabbit Monoclonal Antibody (2Z1K3)

          View all (13) EFTUD2 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite EFTUD2 Recombinant Rabbit Monoclonal Antibody (2Z1K3)

          • Antibody Testing Data (11)
          EFTUD2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          EFTUD2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 11

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          EFTUD2 Antibody (MA5-57936) in ICC/IF

          Immunocytochemistry analysis of EFTUD2 using NIH/3T3 cells. The cells were incubated using 1:400 dilution with EFTUD2 Recombinant Monoclonal Antibody (Product # MA5-57936) (100x lens). Followed by further incubation with Cy3-conjugated Goat anti-Rabbit IgG (H+L) using a 1:500 dilution (Red). DAPI was used for nuclear staining (Blue). Then were counterstained with α-Tubulin Mouse mAb using a 1:400 dilution followed by incubation with ABflo® 488-conjugated Goat Anti-Mouse IgG (H+L) using a 1:500 dilution (Green). DAPI was used for nuclear st... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          EFTUD2 Antibody in Immunocytochemistry (ICC/IF)
          EFTUD2 Antibody in Immunocytochemistry (ICC/IF)
          EFTUD2 Antibody in Immunocytochemistry (ICC/IF)
          EFTUD2 Antibody in Immunocytochemistry (ICC/IF)
          EFTUD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EFTUD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EFTUD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EFTUD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EFTUD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EFTUD2 Antibody in Western Blot (WB)
          EFTUD2 Antibody in Immunoprecipitation (IP)

          Product Details

          MA5-57936

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000-1:30,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:1,000-1:4,000
          -

          Immunocytochemistry (ICC/IF)

          1:200-1:800
          -

          ELISA (ELISA)

          1 µg/mL
          -

          Immunoprecipitation (IP)

          0.5-4 µg for 200-400 µg extracts of whole cells
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          2Z1K3

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 1-205 of human EFTUD2 (NP_004238.3).
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.3, with 0.05% BSA, 50% glycerol

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Dry ice (international)

          RRID

          AB_3680958

          Product Specific Information

          Immunogen sequence: MDTDLYDEFG NYIGPELDSD EDDDELGRET KDLDEMDDDD DDDDVGDHDD DHPGMEVVLH EDKKYYPTAE EVYGPEVETI VQEEDTQPLT EPIIKPVKTK KFTLMEQTLP VTVYEMDFLA DLMDNSELIR NVTLCGHLHH GKTCFVDCLI EQTHPEIRKR YDQDLCYTDI LFTEQERGVG IKSTPVTVVL PDTKGKSYLF NIMDT

          Target Information

          EFTUD2 is component of the U5 snRNP complex required for pre-mRNA splicing.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 116 kDa U5 small nuclear ribonucleoprotein component; Elongation factor Tu GTP-binding domain-containing protein 2; hSNU114; SNU114 homolog; U5 small nuclear ribonucleoprotein 116 kDa; U5 snRNP-specific protein, 116 kDa; U5-116 kDa

          View more View less

          Gene Aliases: 116kDa; EFTUD2; KIAA0031; MFDGA; MFDM; SNRNP116; SNRP116; Snu114; U5-116KD

          View more View less

          UniProt ID: (Human) Q15029, (Mouse) O08810

          View more View less

          Entrez Gene ID: (Human) 9343, (Rat) 287739, (Mouse) 20624

          View more View less

          Function(s)
          GTPase activity protein binding GTP binding poly(A) RNA binding nucleotide binding translation elongation factor
          Process(es)
          mRNA splicing, via spliceosome mRNA processing translation RNA splicing cellular response to drug response to cocaine metabolic process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6ff95d844f-mfs9j:80/100.66.72.166:80.
          git-commit: f44aa9153aef922f8fad9a05e87b8aee811da128
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.28.0-Offline