Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSVPAFIDI SEEDQAAELR AYLKSKGAEI SEENSEGGLH VDLAQIIEAC DVCLKEDDKD VESVMNSVVS LLLILEPDKQ EALIESLCEK LVKFREGERP SLRLQLLSNL FHGMDKNTPV RYTVYCSLIK VAASCGAIQY IPTELDQVRK WISDWNLTTE KKHTLLRLLY EALVDCKKSD AASKVMVELL GSYTEDNASQ ARVDAHRCIV RALKDPNAFL FDHLLTLKPV KFLEGELIHD LLTIFVSAKL ASYVKFYQNN KDFIDSLGLL HEQNMAKMRL LTFMGMAVEN KEISFDTMQQ ELQIGADDVE AFVIDAVRTK MVYCKIDQTQ RKVVVSHSTH RTFGKQQWQQ LYDTLNAWKQ NLNKVKNSLL SLSDT
This gene encodes a protein that is part of the eurkaryotic translation initiation factor 3 complete (eIF-3) required for protein synthesis. Elevated levels of the encoded protein are present in cancer cell lines. Inactivation of the encoded protein has been shown to interfere with translation of herpes virus mRNAs by preventing the association of mRNAs with the ribosomes. A pseudogene of this gene is located on the X chromosome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B5 receptor; dendritic cell protein; dendritic cell protein GA17; EIF3M; Eukaryotic translation initiation factor 3 subunit M; eukaryotic translation initiation factor 3, subunit M; Fetal lung protein B5; FLJ29030; hFL-B5; PCI domain containing 1 (herpesvirus entry mediator); PCI domain-containing protein 1; transport and golgi organization 7 homolog
Gene Aliases: B5; EIF3M; GA17; hfl-B5; HFLB5; PCID1; PNAS-125; RGD1565840; TANGO7
UniProt ID: (Human) Q7L2H7, (Mouse) Q99JX4
Entrez Gene ID: (Human) 10480, (Mouse) 98221, (Rat) 295975
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support