ELF4 is transcriptional activator that binds to DNA sequences containing the consensus 5'-WGGA-3'. The protein acts synergistically with RUNX1 to transactivate the IL3 promoter. Also transactivates the PRF1 promoter in natural killer (NK) cells. This protein plays a role in the development and function of NK and NK T-cells and in innate immunity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E74-like factor 4; E74-like factor 4 (ets domain transcription factor); E74-like factor 4; myeloid elf-1-like factor; myeloid elf-1 like factor; ELF4; ETS-related transcription factor Elf-4; myeloid elf-1 like factor; Myeloid Elf-1-like factor
Gene Aliases: AV314029; BC042423; ELF4; ELFR; ENSMUSG00000053512; Gm9907; MEF; RGD1560743
UniProt ID: (Human) Q99607, (Mouse) Q9Z2U4
Entrez Gene ID: (Human) 2000, (Mouse) 56501, (Rat) 302811
Molecular Function:
nucleic acid binding
signaling molecule
transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support