Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: GNGPHHDRCC TYNENNLVDG VYCLPIGHWI EATGHTNEMK HTTDFYFNIA GHQAMHYSRI LPNIWLGSCP RQVEHVTIKL KHELGITAVM NFQTEWDIV
The Epilepsy, progressive myoclonus type 2A protein (EPM2A) is a dual-specificity phosphatase that associates with polyribosomes. Mutations in this gene have been associated with myoclonic epilepsy of Lafora. EPM2A interacts with a number of proteins known to be involved in glycogen metabolism and has been shown to have robust phosphatase activity against a phosphorylated complex carbohydrate, suggesting that EPM2A may be involved in the regulation of glycogen metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epilepsy, progressive myoclonic epilepsy, type 2 gene alpha; epilepsy, progressive myoclonus type 2, Lafora disease (laforin); EPM2A; Glucan phosphatase; Glycogen phosphatase; Lafora PTPase; Laforin; LAFPTPase; RP1-28C20.2
Gene Aliases: EPM2; EPM2A; MELF
UniProt ID: (Human) O95278, (Rat) Q91XQ2
Entrez Gene ID: (Human) 7957, (Rat) 114005
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support