The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Zebrafish, Bovine and Pig
May have a role in the cell cycle. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Enhancer of rudimentary homolog; Mer; preimplantation protein 1
Gene Aliases: DROER; ERH; Mer; Prei1
UniProt ID: (Human) P84090, (Mouse) P84089
Entrez Gene ID: (Human) 2079, (Mouse) 13877
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support