Product Specifications | |
---|---|
Species Reactivity |
Dog, Human, Mouse, Non-human primate, Rat |
Host/Isotype |
Mouse / IgG2a |
Class |
Monoclonal |
Type |
Antibody |
Clone |
6F8 |
Immunogen |
Protein expressed in 293T cells transfected with human MAPK1 expression vector |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Protein A |
Storage buffer |
PBS with 1mg/mL BSA, 30% glycerol |
Contains |
0.05% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_2536743 |
MA1-099 has been successfully used in Western blot, ChIP, flow cytometry, immunofluorescence, immunohistochemistry and immunoprecipitation applications, and reacts with human, dog, rat, mouse and monkey samples.
MA1-099 has successfully been used in western blot to detect p42 ERK in HeLa, NRK and MDCK cell lysates.
As demonstrated by a Western blot using recombinant ERK1 and ERK2 proteins, MA1-099 can also detect ERK1, but detects ERK2 at much lower concentrations. As demonstrated by a Western blot using a panel of lysates from various species, MA1-099 only detects endogenous ERK2.
ERK2 (also known as MAPK1) is a member of the MAP kinase subfamily of Ser/Thr protein kinases. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. The activation of this kinase requires its phosphorylation by upstream kinases. Upon activation, this kinase translocates to the nucleus of the stimulated cells, where it phosphorylates nuclear targets.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ERK-2; ERT1; Extracellular signal-regulated kinase 2; extracellular-signal-regulated kinase 2; MAP kinase 1; MAP kinase 2; MAP kinase isoform p42; MAPK 1; MAPK 2; mitogen activated protein kinase 1; Mitogen-activated protein kinase 1; Mitogen-activated protein kinase 2; p42-MAPK; protein tyrosine kinase ERK2
Gene Aliases: 9030612K14Rik; AA407128; AU018647; C78273; ERK; ERK-2; ERK2; ERT1; Mapk; MAPK1; MAPK2; p38; p40; p41; p41mapk; p42-MAPK; P42MAPK; PRKM1; PRKM2
UniProt ID: (Human) P28482, (Mouse) P63085, (Rat) P63086
Entrez Gene ID: (Human) 5594, (Dog) 477575, (Mouse) 26413, (Rat) 116590
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support