Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CD361, also known as EVI2B (Ecotropic Viral Integration site 2B) or EVDB, is a poorly characterized type I transmembrane protein, expressed from one of three genes embedded in intron 27b of the neurofibromatosis type 1 (NF1) gene. The DNA strand that is transcribed to produce CD361 is the complementary one to the strand encoding NF1. Murine homolog to human CD361 is associated with ecotropic viral insertions, which have been implicated in the expression of murine myeloid leukemias. CD361 has been also reported to be involved in melanocyte and keratinocyte differentiation. However, it is expressed mainly in peripheral blood and bone marrow.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD361; Ecotropic viral integration site 2B protein homolog; EVI-2B; Protein EVI2B
Gene Aliases: EVDB; EVI2B
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support