Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Endomucin Antibodies

          Invitrogen

          Endomucin Monoclonal Antibody (eBioV.7C7 (V.7C7)), eBioscience™

          Advanced Verification
          47 Published Figures
          67 References
          View all (17) Endomucin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Endomucin Monoclonal Antibody (eBioV.7C7 (V.7C7)), eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          • Published Figures (47)
          • Advanced Verification (1)
          Endomucin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Endomucin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 50

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Endomucin Antibody (14-5851-82) in WB

          Western blot was performed using Anti-Endomucin Monoclonal Antibody (Product # 14-5851-85) and an 80kDa band corresponding to Endomucin was observed across tissue lysates tested except Mouse Bone and Mouse Adipose. Tissue extracts (30 µg lysate) of Mouse Kidney (Lane 1), Mouse Lung (Lane 2), Mouse Bone (Lane 3) and Mouse Adipose (Lane 4) were electrophoresed using Novex® NuPAGE® 4-12 % Bis-Tris gel (Product # NP0322BOX). Resolved proteins were then transferred onto a nitrocellulose membrane (Product # IB23001) by iBlot® 2 Dry Blotting Syst... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Endomucin Antibody in Western Blot (WB)
          Endomucin Antibody in Flow Cytometry (Flow)
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Immunocytochemistry (ICC/IF)
          Endomucin Antibody in Immunocytochemistry (ICC/IF)
          Endomucin Antibody in Immunocytochemistry (ICC/IF)
          Endomucin Antibody in Immunocytochemistry (ICC/IF)
          Endomucin Antibody in Immunohistochemistry, Immunohistochemistry (Frozen), Immunocytochemistry (IHC, IHC (F), ICC/IF)
          Endomucin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Endomucin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Endomucin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Endomucin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Endomucin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Endomucin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Endomucin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Endomucin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Endomucin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Endomucin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Immunohistochemistry (IHC)
          Endomucin Antibody in Flow Cytometry (Flow)
          Endomucin Antibody in Flow Cytometry (Flow)
          Endomucin Antibody in Flow Cytometry (Flow)
          Endomucin Antibody in Flow Cytometry (Flow)
          Endomucin Antibody in Flow Cytometry (Flow)
          Endomucin Antibody in Flow Cytometry (Flow)
          Endomucin Antibody in Flow Cytometry (Flow)
          Endomucin Antibody in Flow Cytometry (Flow)
          Endomucin Antibody in Flow Cytometry (Flow)
          Endomucin Antibody in Flow Cytometry (Flow)
          Endomucin Antibody

          Product Details

          14-5851-82

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 43 publications 43 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-Dependent
          View 5 publications 5 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 8 publications 8 publications

          Flow Cytometry (Flow)

          1 µg/test
          View 6 publications 6 publications

          Immunoprecipitation (IP)

          Assay-Dependent
          -
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eBioV.7C7 (V.7C7)

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 488 
          • eFluor 660
          • PE 
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_891527

          Product Specific Information

          Description: The eBioV.7C7 monoclonal antibody reacts with mouse endomucin, which was identified in a search for cell-surface expressed endothelial cell markers. Endomucin is a 75 kDa type I integral membrane protein, with similarities to the sialomucin family of proteins including extensive O-linked glycosylation. Endomucin is expressed on endothelial cells, however, an exception is the high endothelial venules (HEV) of secondary lymphoid organs. In addition, it has been demonstrated that endomucin is expressed on CD34-c-Kit+Sca-1+Lin- hematopoietic progenitors, and that these cells are capable of multi-lineage long-term reconstitution of the hematopoietic compartment.

          Applications Reported: This eBioV.7C7 (V.7C7) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, immunoblotting (WB), and immunohistology staining of paraffin embedded tissue sections.

          Applications Tested: This eBioV.7C7 (V.7C7) antibody has been tested by flow cytometric analysis of bEnd.3 cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Endomucin (EMCN) also known as endothelial sialomucin or mucin-like sialoglycoprotein interferes with the assembly of focal adhesion complexes and inhibits interaction between cells and the extracellular matrix.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Endomucin; endomucin-1; Endomucin-1/2; MUC-14; mucin 14, endothelial; Mucin-14

          View more View less

          Gene Aliases: 0610012K22Rik; AI315669; Emcn; Muc14

          View more View less

          UniProt ID: (Mouse) Q9R0H2

          View more View less

          Entrez Gene ID: (Mouse) 59308

          View more View less

          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0