Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MKASQCCCCL SHLLASVLLL LLLPELSGPL AVLLQAAEAA PGLGPPDPRP RTLPPLPPGP TPAQQPGRGL AEAAGPRGSE GGNGSNPVAG LETDDHGGKA GEGSVGGGLA VSPNPGDKPM TQRALTVLMV VSGAVLVYFV VRTVRMRRRN RKTRRYGVLD TNIENMELTP LEQDDEDDDN TLFDANHPRR
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 174, member A; HCV NS5A-transactivated protein 6; Hepatitis C virus NS5A-transactivated protein 6; Membrane protein FAM174A; Transmembrane protein 157
Gene Aliases: FAM174A; NS5ATP6; TMEM157; UNQ1912/PRO4371
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support