Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive Control: Siha cell lysate, THP-1 cell lysate, human colon carcinoma tissue.
Subcellular Location: Cell membrane, Endoplasmic reticulum, Membrane.
Tethers the endoplasmic reticulum to the cell membrane and promotes the formation of appositions between the endoplasmic reticulum and the cell membrane. Binds glycerophospholipids in a barrel-like domain and may play a role in cellular lipid transport. Plays a role in FGF signaling via its role in the rapid internalization of FGFR1 that has been activated by FGF1 binding; this occurs most likely via the AP-2 complex. Promotes the localization of SACM1L at endoplasmic reticulum-plasma membrane contact sites (EPCS) (PubMed:27044890). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chr2 synaptotagmin; Chr2Syt; E-Syt2; extended synaptotagmin like protein 2; extended synaptotagmin protein 2; Extended synaptotagmin-2; extended synaptotagmin-like protein 2; family with sequence similarity 62 (C2 domain containing), member B
Gene Aliases: CHR2SYT; E-Syt2; ESYT2; FAM62B; KIAA1228
UniProt ID: (Human) A0FGR8
Entrez Gene ID: (Human) 57488
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support