Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FAM98A belongs to the FAM98 family. The function of FAM98A remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 98, member A; Protein FAM98A
Gene Aliases: FAM98A
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support