Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FBXO5 Antibodies

          Invitrogen

          FBXO5 Recombinant Rabbit Monoclonal Antibody (9F9)

          View all (12) FBXO5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite FBXO5 Recombinant Rabbit Monoclonal Antibody (9F9)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (14)
          FBXO5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FBXO5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 14

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FBXO5 Antibody (MA572992) in ICC/IF

          Immunocytochemistry analysis of FBXO5 in HeLa cells. Permeabilized with Trinton X-100 at room temp for 20 min. Samples were incubated with FBXO5 Recombinant Monoclonal (Product # MA572992 using a dilution at 1:100, for 90 min at 37°C. Followed by conjugated Goat Anti-Rabbit IgG antibody (green) at 37°C for 90 min, DAPI was used to stain the cell nuclei. PBS instead of the primary antibody was used as the blank control. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FBXO5 Antibody in Immunocytochemistry (ICC/IF)
          FBXO5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO5 Antibody in Western Blot (WB)
          FBXO5 Antibody in Flow Cytometry (Flow)

          Product Details

          MA572992

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:100
          -

          Flow Cytometry (Flow)

          1:50-1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          9F9

          Immunogen

          KLH conjugated synthetic peptide derived from human FBXO5.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.175 mg/mL

          Amount

          17.5 µg

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4, with 50% glycerol, 1% BSA

          Contains

          0.02% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          F-box proteins, critical components of the conserved ubiquitin-protein ligase complex SCF (Skp1/Cdc53-Cullin1/F-box), recruit substrates for ubiquitination and consequent degradation through their specific protein-protein interaction domains. Novel human F-box proteins named FBG1, FBG2, FBG3, FBG4 and FBG5 constitute a subfamily of mammalian F-box proteins. FBG genes are expressed in a limited number of human tissues including kidney, liver, brain and muscle tissues.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: early fission inhibitory protein 1; Early mitotic inhibitor 1; f-box only protein 31; F-box only protein 5; F-box protein Fbx5; fbxo5; unnamed protein product

          View more View less

          Gene Aliases: 2510044I10Rik; C85305; EMI1; FBX5; Fbxo31; FBXO5

          View more View less

          UniProt ID: (Human) Q9UKT4, (Mouse) Q7TSG3

          View more View less

          Entrez Gene ID: (Human) 26271, (Mouse) 67141, (Rat) 292263

          View more View less

          Function(s)
          protein binding zinc ion binding anaphase-promoting complex binding protein kinase binding metal ion binding molecular function inhibitor activity ubiquitin ligase inhibitor activity
          Process(es)
          oocyte maturation cell cycle spindle assembly involved in female meiosis I mitotic nuclear division regulation of mitotic cell cycle vesicle organization regulation of meiotic nuclear division negative regulation of meiotic nuclear division microtubule polymerization spindle assembly cell division negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle negative regulation of ubiquitin-protein ligase activity involved in meiotic cell cycle positive regulation of meiosis I obsolete inhibition of APC-Cdc20 complex activity regulation of DNA replication cellular response to DNA damage stimulus regulation of mitotic nuclear division positive regulation of cell proliferation negative regulation of cell cycle process positive regulation of G2/M transition of mitotic cell cycle protein ubiquitination negative regulation of DNA endoreduplication positive regulation of osteoblast differentiation negative regulation of mitotic metaphase/anaphase transition negative regulation of ubiquitin-protein transferase activity positive regulation of biomineral tissue development negative regulation of ubiquitin protein ligase activity positive regulation of mesenchymal stem cell migration negative regulation of cellular senescence
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6479c7f679-2z78h:80/100.66.76.236:80.
          git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.50.0-Offline