Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive Samples: Mouse heart, Rat heart, SKOV3, RD
Immunogen sequence: DCKDLSYKDR HWHEACFHCS QCRNSLVDKP FAAKEDQLLC TDCYSNEYSS KCQECKKTIM PGTRKMEYKG SSWHETCFIC HRCQQPIGTK SFIPKDNQNF CVPCYEKQHA MQCVQCKKPI TTGGVTYREQ PWHKECFVCT ACRKQLSGQR FTARDDFAYC LNCFCDLYAK KCAGCTNPIS GLGGTKYISF EERQWHNDCF NCKKCSLSLV G
This gene encodes a member of the four-and-a-half-LIM-only protein family. Family members contain two highly conserved, tandemly arranged, zinc finger domains with four highly conserved cysteines binding a zinc atom in each zinc finger. This protein is thought to have a role in the assembly of extracellular membranes. Also, this gene is down-regulated during transformation of normal myoblasts to rhabdomyosarcoma cells and the encoded protein may function as a link between presenilin-2 and an intracellular signaling pathway. Multiple alternatively spliced variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aging-associated gene 11; down-regulated in rhabdomyosarcoma LIM protein; FHL-2; Four and a half LIM domains protein 2; LIM domain protein DRAL; OTTHUMP00000203164; OTTHUMP00000203168; OTTHUMP00000203169; Skeletal muscle LIM-protein 3; SLIM 3; SLIM-3
Gene Aliases: AAG11; DRAL; FHL-2; FHL2; SLIM-3; SLIM3
UniProt ID: (Human) Q14192, (Mouse) O70433, (Rat) O35115
Entrez Gene ID: (Human) 2274, (Mouse) 14200, (Rat) 63839
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support