Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MALYELFSHP VERSYRAGLC SKAALFLLLA AALTYIPPLL VAFRSHGFWL KRSSYEEQPT VRFQHQVLLV ALLGPESDGF LAWSTFPAFN RLQGDRLRVP LVSTREEDRN QDGKTDMLHF KLELPLQSTE HVLGVQLILT FSYRLHRMAT LVMQSMAFLQ SSFPVPGSQL YVNGDLRLQQ KQPLSCGGLD ARYNISVING TSPFAYDYDL THIVAAYQER NVTTVLNDPN PIWLVGRAAD APFVINAIIR YPVEVISYQP GFWEMVKFAW VQYVSILLIF LWVFERIKIF VFQNQVVTTI PVTVTPRGDL CKEHLS
The function remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Transmembrane protein 231
Gene Aliases: ALYE870; JBTS20; MKS11; PRO1886; TMEM231; UNQ870/PRO1886
UniProt ID: (Human) Q9H6L2
Entrez Gene ID: (Human) 79583
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support