Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MDLTKREYTM HDHNTLLKEF ILNNEGKLKK LQDDAKIAQD AFDDVVKYFG ENPKTTPPSV FFPVFVRFVK AYKQAEEENE LRKKQEQALM EKLLEQEALM EQQDPKSPSH KSKRQQQELI AELRRRQVKD NRHVYEGKDG AIEDIITALK KNNITKFPNV HSRVRISSST PVVEDTQS
Formin-like protein 2 is a cytoplasmic phosphoprotein belonging to the formin homology domain containing protein family and has a DAD (diaphanous autoregulatory domain), FH2 (formin homology 2) domain, and GBD/FH3 (Rho GTPase-binding/formin homology 3) domain. The exact function of FMNL2 is yet to be known but reports suggest that it may have a role in the Wnt signaling pathway. Formin-related proteins are also known to play an important role in morphogenesis, cytokinesis and cell polarity. Several alternatively spliced transcript variants have been described but their full-length nature is not yet determined. It is also known to be associated with a diffuse-type gastric cancer, breast cancer, chondrosarcoma, melanoma, and glioblastoma. FMNL2 might be implicated in polarity control, invasion, migration or metastasis through actin filament nucleation and may be regulated by Rho family GTPase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: formin homology 2 domain containing 2; Formin homology 2 domain-containing protein 2; formin-like 2; Formin-like protein 2
Gene Aliases: FHOD2; FMNL2; KIAA1902
UniProt ID: (Human) Q96PY5
Entrez Gene ID: (Human) 114793
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support