Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000 | - |
Immunohistochemistry (IHC) |
1:50-1:200 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Between 329 and 379 |
Conjugate |
Unconjugated |
Form |
liquid |
Concentration |
0.10 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
TBS with 0.1% BSA |
Contains |
0.09% sodium azide |
Storage conditions |
4° C |
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
FNTA (Farnesyltransferase, CAAX box; alphaCAAX farnesyltransferase alpha subunit, FPTA, FTase-alpha, GGTase-I-alpha, Protein farnesyl transferase/ geranyl geranyl transferase type I alpha subunit(PGGT1A), Ras proteins prenyl transferase alpha, Type I protein geranyl-geranyl transferase alpha subunit) is located locus 8p11 (human). Prenylation is a common intracellular posttranslational protein modification catalyzed by prenyltransferases. The best characterized among these prenyltransferases is the CAAX farnesyl transferase (2. 5. 1. 21), which is alpha-beta heterodimeric enzyme. It catalyzes the attachment of a farnesyl group from farnesyl phosphate to cysteine residues at the fourth position from the C terminus of proteins that end in the CAAX box. FNTA or the alpha subunit of farnesyl transferase is shared by the geranyl-geranyl protein transferases. Apart from transfer of farnesyl and geranyl groups to proteins, FNTA has been proposed to regulate intracellular signaling pathways involving p21ras. Reports have shown that FNTA interacts with TGF-beta and activin-type I receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAAX farnesyltransferase subunit alpha; farnesyl-protein transferase alpha-subunit; FTase-alpha; GGTase-I-alpha; Protein farnesyltransferase/geranylgeranyltransferase type-1 subunit alpha; protein prenyltransferase alpha subunit repeat containing 2; Ras proteins prenyltransferase subunit alpha; type I protein geranyl-geranyltransferase alpha subunit; Type I protein geranyl-geranyltransferase subunit alpha
Gene Aliases: FNTA; FPTA; PGGT1A; PTAR2
UniProt ID: (Human) P49354
Entrez Gene ID: (Human) 2339
Molecular Function:
acyltransferase
viral or transposable element protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support