Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FOXC2 Antibodies

          Proteintech

          FOXC2 Recombinant Rabbit Monoclonal Antibody (240403H7)

          View all (18) FOXC2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite FOXC2 Recombinant Rabbit Monoclonal Antibody (240403H7)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (4)
          FOXC2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FOXC2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FOXC2 Antibody (83476-5-RR) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed A375 cells using FOXC2 antibody (83476-5-RR, Clone: 240403H7) at dilution of 1:300 and CoraLite®488-Conjugated Goat Anti-Mouse IgG (H+L) (SA00013-1), CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FOXC2 Antibody in Immunocytochemistry (ICC/IF)
          FOXC2 Antibody in Flow Cytometry (Flow)
          FOXC2 Antibody in Flow Cytometry (Flow)
          FOXC2 Antibody in Surface plasmon resonance (SPR)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          FOXC2 Recombinant Rabbit Monoclonal Antibody (240403H7)

          Product Details

          83476-5-RR

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1:150-1:600
          -

          Flow Cytometry (Flow)

          0.25 µg/10^6 cells in a 100 µL suspension
          -

          ELISA (ELISA)

          Assay-dependent
          -

          Surface plasmon resonance (SPR)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293F

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          240403H7

          Immunogen

          FOXC2 fusion protein Ag33121
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: KEAEKKVVIK SEAASPALPV ITKVETLSPE SALQGSPRSA ASTPAGSPDG SLPEHHAAAP NGLPGFSVEN IMTLRTSPPG GELSPGAGRA GLVVPPLALP YAAAPPAAYG QPCAQGLEAG AAGGYQCSMR AMSLYTGAER PAHMCVPPAL DEALSDHPSG PTSPLSALNL AAGQEGALAA TGHHHQHHGH HHPQAPPPPP APQPQPTPQP GAAAAQAASW YLNHSGDLNH LPGHTFAAQQ QTFPNVREMF NSHRLGIENS TLGESQVSGN ASCQLPYRST PPLYRHAAPY SYDCTKY

          Target Information

          Forkhead box protein C2 (FOXC2) is a transciptional activator and is characterized by a distinct DNA-binding forkhead domain. FOXC2 may be involved in the formation of special mesenchymal tissues. Mutations affecting the gene can cause lymphedema-distichiasis (LYD).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: forkhead box C2 (MFH-1, mesenchyme forkhead 1); forkhead, Drosophila, homolog-like 14; forkhead-related protein FKHL14; mesenchyme fork head protein 1; mesenchyme forkhead 1; MFH-1,mesenchyme forkhead 1; transcription factor FKH-14

          View more View less

          Gene Aliases: FKHL14; LD; MFH-1; MFH1

          View more View less

          UniProt ID: (Human) Q99958

          View more View less

          Entrez Gene ID: (Human) 2303

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding bacterial-type RNA polymerase transcriptional activator activity, sequence-specific DNA binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding chromatin DNA binding identical protein binding sequence-specific DNA binding sequence-specific double-stranded DNA binding promoter-specific chromatin binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter somitogenesis lymphangiogenesis regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter mesoderm development heart development insulin receptor signaling pathway anatomical structure morphogenesis response to hormone positive regulation of endothelial cell migration cell differentiation positive regulation of cell adhesion mediated by integrin positive regulation of vascular wound healing positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of cell migration involved in sprouting angiogenesis negative regulation of cold-induced thermogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-wx6nt:80/100.66.72.150:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0