Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a member of the forkhead/winged helix-box (FOX) family of transcription factors. FOX transcription factors play critical roles in the regulation of multiple processes including metabolism, cell proliferation and gene expression during ontogenesis. Mutations in this gene are associated with a complex phenotype consisting of dilated cardiomyopathy, obsessive-compulsive disorders, and suicidality.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: forkhead box D4 like 3; forkhead box D4-like 2; Forkhead box protein D4; Forkhead box protein D4-like 2; Forkhead box protein D4-like 3; Forkhead box protein D4-like 4; Forkhead box protein D4-like 5; Forkhead box protein D4-like 6; Forkhead box protein D4B; forkhead, Drosophila, homolog-like 9; forkhead-related activator 5; Forkhead-related protein FKHL9; Forkhead-related transcription factor 5; FOXD4-like 2; FOXD4-like 3; FOXD4-like 4; FOXD4-like 5; FOXD4-like 6; FREAC-5; Myeloid factor-alpha; Myeloid factor-gamma; winged helix factor-1; winged helix transcription factor beta
Gene Aliases: FKHL9; FOXD4; FOXD4A; FOXD4B; FOXD4L2; FOXD4L3; FOXD4L4; FOXD4L5; FOXD4L6; FREAC5
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support