Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FXR1 Antibodies

          Invitrogen

          FXR1 Recombinant Rabbit Monoclonal Antibody (9D20)

          View all (21) FXR1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite FXR1 Recombinant Rabbit Monoclonal Antibody (9D20)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (16)
          FXR1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FXR1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 16

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FXR1 Antibody (MA573169) in ICC/IF

          Immunocytochemistry analysis of FXR1 in MCF-7 cells. Permeabilized with Trinton X-100 at room temp for 20 min. Samples were incubated with FXR1 Recombinant Monoclonal (Product # MA573169 using a dilution at 1:100, for 90 min at 37°C. Followed by conjugated Goat Anti-Rabbit IgG antibody (green) at 37°C for 90 min, DAPI was used to stain the cell nuclei. PBS instead of the primary antibody was used as the blank control. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FXR1 Antibody in Immunocytochemistry (ICC/IF)
          FXR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FXR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FXR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FXR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FXR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FXR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FXR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FXR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FXR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FXR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FXR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FXR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FXR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FXR1 Antibody in Western Blot (WB)
          FXR1 Antibody in Flow Cytometry (Flow)

          Product Details

          MA573169

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:500
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          Flow Cytometry (Flow)

          1:50-1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          9D20

          Immunogen

          KLH conjugated synthetic peptide derived from human FXR1.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4, with 50% glycerol, 1% BSA

          Contains

          0.02% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          FXR1 is an RNA binding protein that interacts with the functionally-similar proteins FMR1 and FXR2. It may regulate intracellular transport and local transltion of certain mRNAs. These proteins shuttle between the nucleus and cytoplasm and associate with polyribosomes, predominantly with the 60S ribosomal subunit. FXR1 is required for embryonic and postnatal development of muscle tissue. Three transcript variants encoding different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: FMR1 autosomal homolog 1; FMR1 autosomal protein-like protein 1; fragile X mental retardation gene, autosomal homolog; fragile X mental retardation syndrome-related protein 1; fragile X mental retardation-related protein 1; fragile-X-related protein 1; FXR1P; hFXR1p; mFxr1p; RNA-binding protein FXR1; unnamed protein product

          View more View less

          Gene Aliases: 1110050J02Rik; 9530073J07Rik; AI851072; CMYO9A; CMYO9B; CMYP9A; CMYP9B; FXR1; Fxr1h; FXR1P; MYOPMIL; MYORIBF

          View more View less

          UniProt ID: (Human) P51114, (Rat) Q5XI81, (Mouse) Q61584

          View more View less

          Entrez Gene ID: (Human) 8087, (Rat) 361927, (Mouse) 14359

          View more View less

          Function(s)
          nucleic acid binding RNA binding mRNA binding mRNA 3'-UTR binding protein binding RNA strand annealing activity mRNA 3'-UTR AU-rich region binding protein homodimerization activity ribonucleoprotein complex binding translation regulator activity protein heterodimerization activity molecular condensate scaffold activity G-quadruplex RNA binding poly(A) RNA binding translation factor
          Process(es)
          cytoplasmic translational initiation regulation of translation apoptotic process spermatogenesis spermatid development muscle organ development posttranscriptional regulation of gene expression negative regulation of translation dentate gyrus development cell differentiation negative regulation of tumor necrosis factor production positive regulation of Rho protein signal transduction regulation of mRNA stability regulation of circadian sleep/wake cycle, sleep positive regulation of translation positive regulation of long-term neuronal synaptic plasticity animal organ development negative regulation of inflammatory response regulation of neurogenesis mRNA transport nuclear pore complex assembly nuclear pore localization regulation of synaptic transmission, glutamatergic skeletal muscle organ development muscle structure development mRNA destabilization regulation of translation at presynapse, modulating synaptic transmission membraneless organelle assembly negative regulation of long-term synaptic potentiation negative regulation of mRNA catabolic process positive regulation of gene silencing by miRNA multicellular organism development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6479c7f679-74pjh:80/100.66.78.195:80.
          git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.50.0-Offline