Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MASLGHILV FCVGLLTMAK AESPKEHDPF TYDYQSLQIG GLVIAGILFI LGILIVLSRR CRCKFNQQQR TGEPDEEEGT FRSSIRRLST RRR
This gene encodes a member of the FXYD family of small membrane proteins that share a 35-amino acid signature sequence domain, beginning with the sequence PFXYD and containing 7 invariant and 6 highly conserved amino acids. The protein encoded by this gene is a plasma membrane substrate for several kinases, including protein kinase A, protein kinase C, NIMA kinase, and myotonic dystrophy kinase. It is thought to form an ion channel or regulate ion channel activity and act as an accessory protein of Na,K-ATPase. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FXYD domain-containing ion transport regulator 1; Phospholemman; Sodium Potassium ATPase; Sodium/potassium-transporting ATPase subunit FXYD1
Gene Aliases: 0610012C17Rik; 1110006M24Rik; FXYD1; PLM; Pml
UniProt ID: (Human) O00168, (Mouse) Q9Z239
Entrez Gene ID: (Human) 5348, (Mouse) 56188
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support