Product References
Association of Fascin and matrix metalloproteinase-9 expression with poor prognostic parameters in breast carcinoma of Egyptian women.
Diagnostic pathology
Youssef NS,Hakim SA
Published figure using Fascin monoclonal antibody (Product # MA5-11483) in Immunohistochemistry
Fri Jul 04 00:00:00 EDT 2014
Association of Fascin and matrix metalloproteinase-9 expression with poor prognostic parameters in breast carcinoma of Egyptian women.
Diagnostic pathology
Youssef NS,Hakim SA
Published figure using Fascin monoclonal antibody (Product # MA5-11483) in Immunohistochemistry
Fri Jul 04 00:00:00 EDT 2014
The correlation of cortactin and fascin-1 expression with clinicopathological parameters in pancreatic and ampulla of Vater adenocarcinoma.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica
Tsai WC,Lin CK,Lee HS,Gao HW,Nieh S,Chan DC,Jin JS
MA5-11483 was used in immunohistochemistry to study the clinicopathology of pancreatic and ampulla of Vater adenocarcinomas and the corresponding expression of cortactin and fascin-1
Fri Mar 01 00:00:00 EST 2013
EGFR signaling regulates tumor cell migration in craniopharyngiomas.
Clinical cancer research : an official journal of the American Association for Cancer Research
Hölsken A,Gebhardt M,Buchfelder M,Fahlbusch R,Blümcke I,Buslei R
MA5-11483 was used in immunohistochemistry and western blot to study the role of EGFR signaling in regulating tumor cell migration in craniopharyngiomas
Fri Jul 01 00:00:00 EDT 2011
Fascin expression and its potential significance in gastrointestinal stromal tumors.
The Turkish journal of gastroenterology : the official journal of Turkish Society of Gastroenterology
Ozcan A,Karslioğlu Y,Günal A,Cermık AH,Kurt B,Ongürü O
MA5-11483 was used in immunohistochemistry to study the expression of fascin in gastrointestinal stromal tumors and its possible prognostic value
Mon Aug 01 00:00:00 EDT 2011
Prognostic significance of fascin-1 and E-cadherin expression in laryngeal squamous cell carcinoma.
European journal of cancer prevention : the official journal of the European Cancer Prevention Organisation (ECP)
Zou J,Yang H,Chen F,Zhao H,Lin P,Zhang J,Ye H,Wang L,Liu S
MA5-11483 was used in immunohistochemistry to study the prognostic significance of fascin-1 and E-cadherin expression in laryngeal squamous cell carcinoma
Fri Jan 01 00:00:00 EST 2010
Availability of activated CD4+ T cells dictates the level of viremia in naturally SIV-infected sooty mangabeys.
The Journal of clinical investigation
Klatt NR,Villinger F,Bostik P,Gordon SN,Pereira L,Engram JC,Mayne A,Dunham RM,Lawson B,Ratcliffe SJ,Sodora DL,Else J,Reimann K,Staprans SI,Haase AT,Estes JD,Silvestri G,Ansari AA
MA5-11483 was used in immunohistochemistry to study the role of CD4+ T-cells in determining the viral load set-point in sooty mangabeys naturally infected with SIV
Sun Jun 01 00:00:00 EDT 2008
Proteomic identification of novel proteins in cortical lewy bodies.
Brain pathology (Zurich, Switzerland)
Leverenz JB,Umar I,Wang Q,Montine TJ,McMillan PJ,Tsuang DW,Jin J,Pan C,Shin J,Zhu D,Zhang J
MA5-11483 was used in immunohistochemistry to identify novel proteins in cortical lewy bodies using proteomics
Sun Apr 01 00:00:00 EDT 2007