Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: RQKRSIHEPD PTAVEQISLE SVDMDSPVNM KFNLSHLGSK EHILELRPAI QPLNNHIRYV ISQGNDDSVF RIHQRNGLSY LHTAKKKLMP GTYTLEITSI P
The protein encoded by this gene is a component of connective tissue microfibrils and may be involved in elastic fiber assembly. Mutations in this gene cause congenital contractural arachnodactyly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fibrillin 5; Fibrillin-2; Placensin
Gene Aliases: CCA; DA9; EOMD; FBN2
UniProt ID: (Human) P35556
Entrez Gene ID: (Human) 2201
Molecular Function: extracellular matrix structural protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support