Invitrogen
Applications | Tested Dilution | Publications |
---|---|---|
Immunohistochemistry (IHC) |
Assay-dependent | - |
Immunocytochemistry (ICC/IF) |
Assay-dependent | View 2 publications 2 publications |
Flow Cytometry (Flow) |
Assay-dependent | View 1 publication 1 publication |
Miscellaneous PubMed (Misc) |
- | View 2 publications 2 publications |
Product Specifications | |
---|---|
Species Reactivity |
Chemical |
Published species |
Not Applicable |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Fluorescein/Oregon Green |
Conjugate |
Biotin |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
purified |
Storage buffer |
0.1M potassium phosphate, pH 8 |
Contains |
5mM sodium azide |
Storage conditions |
4° C |
RRID |
AB_2536213 |
Anti-fluorescent dye antibodies recognize specific fluorophores and, in most cases, quench their fluorescence. Thus many anti-dye antibodies, including those that recognize fluorescein, can serve as cell-impermeant probes for determining whether fluorescent dye-conjugated ligands, proteins, bacteria or other biomolecules have been internalized by endocytic or pinocytic processes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FITC
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support